Skip to Content
Merck
All Photos(1)

Key Documents

S5068

Sigma-Aldrich

Salmeterol xinafoate

≥98% (HPLC), solid

Synonym(s):

(±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: slightly soluble
ethanol: slightly soluble
methanol: freely soluble

originator

GlaxoSmithKline

SMILES string

OC(=O)c1ccc2ccccc2c1O.OCc3cc(ccc3O)C(O)CNCCCCCCOCCCCc4ccccc4

InChI

1S/C25H37NO4.C11H8O3/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21;12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2;1-6,12H,(H,13,14)

InChI key

XTZNCVSCVHTPAI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

Application

Salmeterol xinafoate was used as conjugate with human serum albumin to study the immune responses in rabbits.7

Biochem/physiol Actions

β2-adrenoceptor agonist.
Salmeterol xinafoate is a β2 adrenoceptor agonist with longer lasting bronchodilation and anti-inflammatory effects. It relaxes the airway smooth muscles, stabilizes mast cells and regulates the release of histamine. Salmetrol is particularly effective in treatment of mild asthma, chronic obstructive pulmonary disorder.5,6

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Slack et al.
The Journal of pharmacology and experimental therapeutics, 344(1), 218-230 (2012-11-08)
Vilanterol trifenatate (vilanterol) is a novel, long-acting β(2)-adrenoceptor (β(2)-AR) agonist with 24 h activity. In this study, we describe the preclinical pharmacological profile of vilanterol using radioligand binding and cAMP studies in recombinant assays as well as human and guinea
Terence L Fodey et al.
Analytica chimica acta, 637(1-2), 328-332 (2009-03-17)
Haptens are low molecular weight compounds that are non-immunogenic and so must be conjugated to carrier molecules to elicit an immune response. Doses of 50-1000 microg protein conjugate have been suggested for immunisation of rabbits with hapten-protein immunogens. Although larger
Basak Isin et al.
PloS one, 7(12), e50186-e50186 (2013-01-10)
Recently available G-protein coupled receptor (GPCR) structures and biophysical studies suggest that the difference between the effects of various agonists and antagonists cannot be explained by single structures alone, but rather that the conformational ensembles of the proteins need to
M Isogaya et al.
Molecular pharmacology, 54(4), 616-622 (1998-10-10)
Transmembrane domains (TMDs) I, II, and VII of the beta2-adrenergic receptor (beta2AR) were replaced, individually or in combination, with the corresponding regions of the beta1AR, and vice versa. The beta2-selective binding of salmeterol was not affected by the exchange of
Marc Decramer et al.
Respiratory medicine, 107(2), 223-232 (2012-12-12)
Indacaterol is a once-daily, long-acting β(2)-agonist bronchodilator that improves dyspnoea and health status in patients with moderate-to-severe COPD. While its bronchodilator effects have been shown to be maintained in different patient subgroups, effects on clinical outcomes in certain subgroups are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service