Skip to Content
Merck
All Photos(1)

Documents

109630

Sigma-Aldrich

1-Hydroxy-2-naphthoic acid

≥97.0%

Synonym(s):

α-Hydroxynaphthoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6CO2H
CAS Number:
Molecular Weight:
188.18
Beilstein:
974438
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0%

mp

195-200 °C (dec.) (lit.)

solubility

alcohol: freely soluble
benzene: freely soluble
cold water: insoluble
diethyl ether: freely soluble

SMILES string

OC(=O)c1ccc2ccccc2c1O

InChI

1S/C11H8O3/c12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h1-6,12H,(H,13,14)

InChI key

SJJCQDRGABAVBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hydroxy-2-naphthoic acid is produced by the transformation of phenanthrene by NCIB 9816 clones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Hanna et al.
Chemosphere, 70(2), 178-186 (2007-08-11)
The adsorption of naphthoic acids to iron oxides and hydroxides influences strongly their mobility in soils and sediments. Sorption of 1-hydroxy-2-naphthoic acid (HNA) to three iron oxides was examined over a wide range of conditions (pH, ionic strength, sorbate and
Marta Ferraroni et al.
Journal of structural biology, 177(2), 431-438 (2011-12-14)
The crystallographic structures of the adducts of salicylate 1,2-dioxygenase (SDO) with substrates salicylate, gentisate and 1-hydroxy-2-naphthoate, obtained under anaerobic conditions, have been solved and analyzed. This ring fission dioxygenase from the naphthalenesulfonate-degrading bacterium Pseudaminobacter salicylatoxidans BN12, is a homo-tetrameric class
Marta Ferraroni et al.
The FEBS journal, 280(7), 1643-1652 (2013-02-07)
The salicylate 1,2-dioxygenase (SDO) from the bacterium Pseudaminobacter salicylatoxidans BN12 is a versatile gentisate 1,2-dioxygenase (GDO) that converts both gentisate (2,5-dihydroxybenzoate) and various monohydroxylated substrates. Several variants of this enzyme were rationally designed based on the previously determined enzyme structure
Mario Cazzola et al.
Clinical pharmacokinetics, 41(1), 19-30 (2002-02-05)
Salmeterol is an inhaled long-acting selective beta(2)-adrenoceptor agonist that is commercially available as the xinafoate (1-hydroxy-2-naphthoic acid) salt of the racemic mixture of the two optical isomers, (R)- and (S)-, of salmeterol. It acts locally in the lung through action
Tony Hadibarata et al.
Journal of environmental management, 111, 115-119 (2012-07-28)
The biodegradation of benzo[a]pyrene (BaP) by using Polyporus sp. S133, a white-rot fungus isolated from oil-contaminated soil was investigated. Approximately 73% of the initial concentration of BaP was degraded within 30 d of incubation. The isolation and characterization of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service