Skip to Content
Merck
All Photos(1)

Documents

S3917

Sigma-Aldrich

Single-strand Binding Protein from Escherichia coli

>95% (SDS-PAGE), for molecular biology, Single strand binding protein to enhance PCR specificity

Synonym(s):

SSB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.52

grade

for molecular biology

Quality Level

Assay

>95% (SDS-PAGE)

form

liquid

color

colorless

foreign activity

DNase and RNase, none detected

shipped in

wet ice

storage temp.

−20°C

General description

Single-strand Binding Protein is a homotetramer, found in E. coli during the event of DNA metabolism.

Application

Binds with high specificity to single-stranded DNA. Can be useful in enhancing the specificity of PCR and in enabling the sequencing of problematic DNA templates.
Single-strand Binding Protein from Escherichia coli has been used in in vitro RNA/Single-Strand Binding Protein (SSB) complex formation assays and in protein purification to determine integrity and functionality of the integron attC sites.

Biochem/physiol Actions

Single-strand Binding Protein (SSBP) exhibits high affinity towards single stranded DNA important for DNA metabolic processes. Single-stranded DNA-binding protein hSSB1 is critical for genomic stability and assists the delivery of primers during the formation of lagging standard. It also prevents the formation of inhibitory secondary structures in the exposed lagging-strand of DNA during replication. SSBP mediates the recruitment of other proteins to their sites of function on DNA.

Physical form

Solution in 20 mM Tris-HCl, pH 8.0, 0.5 M NaCl, 0.1 mM EDTA, 0.1 mM DTT, 50% glycerol

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Translation, but not transfection limits clinically relevant, exogenous mRNA based induction of alpha-4 integrin expression on human mesenchymal stem cells
Nowakowski A, et al.
Scientific reports, 7(1103), 1-12 (2017)
Single-stranded DNA-binding protein hSSB1 is critical for genomic stability
Richard DJ, et al.
Nature, 453(7195), 677-677 (2008)
Structure of the SSB--DNA polymerase III interface and its role in DNA replication
Marceau AH, et al.
The Embo Journal, 30(20), 4236-4247 (2011)
Binding Specificity of E. coli SSB protein for the ? subunit of DNA pol III Holoenzyme and PriA helicase
Alexander GK, et al.
Biochemistry, 49(17), 3555-3556 (2010)
Minimizing deletion mutagenesis artifact during Taq DNA polymerase PCR by E. coli SSB.
Q Chou
Nucleic acids research, 20(16), 4371-4371 (1992-08-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service