Skip to Content
Merck
All Photos(4)

Key Documents

P7936

Sigma-Aldrich

Phosphocreatine disodium salt hydrate

≥97%

Synonym(s):

Creatine phosphate disodium salt, N-(Imino[phosphonoamino]methyl)-N-methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8N3O5PNa2 · x H2O
CAS Number:
Molecular Weight:
255.08 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Phosphocreatine disodium salt hydrate, ≥97%

Assay

≥97%

form

powder

impurities

≤0.02% free creatine

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].CN(CC(O)=O)C(=N)NP([O-])([O-])=O

InChI

1S/C4H10N3O5P.2Na.H2O/c1-7(2-3(8)9)4(5)6-13(10,11)12;;;/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);;;1H2/q;2*+1;/p-2

InChI key

KZVYPOKBYRXZKS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Phosphocreatine disodium salt hydrate has been used:
  • as a component: of Xenopus buffer (XB) buffer to remove the membrane-bound proteins
  • of M5+ buffer to perform polarized total internal reflection fluorescence (polTIRF) experiments
  • of ATP regenerating system for 7-nitro-2-1,3-benzoxadiazol-4-yl phospholipid(NBD-PL) translocation assay

Biochem/physiol Actions

Phosphocreatine disodium salt hydrate is useful to study nasopharyngeal carcinoma cells and induced temporary cardiac arrest. It also evaluate the effects of adenosine A(2A) receptor modulation in the nucleus accumbens on cocaine seeking in rats.
Phosphocreatine is widely known for its quickest form of regeneration using enzyme creatine kinase. It functions as a temporal energy buffer. Phosphocreatine is present in muscle tissues and plays a major role in maintaining energy homeostasis in muscle cells.
Phosphocreatine, a high-energy phosphate reservoir in vertebrate and some invertebrate muscles, provides phosphate for ADP-ATP conversion.

Preparation Note

Prepared enzymatically

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Guimarães-Ferreira
Einstein (Sao Paulo, Brazil), 12(1), 126-131 (2014-04-15)
Adenosine triphosphate is the present energy currency in the body, and is used in various cellular and indispensable processes for the maintenance of cell homeostasis. The regeneration mechanisms of adenosine triphosphate, from the product of its hydrolysis - adenosine diphosphate
Casey E O'Neill et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 37(5), 1245-1256 (2011-12-16)
Repeated cocaine administration enhances dopamine D(2) receptor sensitivity in the mesolimbic dopamine system, which contributes to drug relapse. Adenosine A(2A) receptors are colocalized with D(2) receptors on nucleus accumbens (NAc) medium spiny neurons where they antagonize D(2) receptor activity. Thus
Protective effects of a magnesium magnetic isotope (Mg(25) )-exchanging nanoparticle ((25) MgPMC(16) ) on mitochondrial functional disorders in esmolol-induced cardiac arrest in rats
Adeli, S., et al.
Auton. Autocoid Pharmacol., 32, 1-7 (2012)
Cyril Bernis et al.
Methods in cell biology, 122, 165-191 (2014-05-27)
The large and complex eukaryotic nucleus is the arbiter of DNA replication, RNA transcription, splicing, and ribosome assembly. With the advent of in vitro nuclear reconstitution extracts derived from Xenopus eggs in the 1980s, it became possible to assemble multiple
John F Beausang et al.
Biophysical journal, 104(6), 1263-1273 (2013-03-27)
Myosin V is biomolecular motor with two actin-binding domains (heads) that take multiple steps along actin by a hand-over-hand mechanism. We used high-speed polarized total internal reflection fluorescence (polTIRF) microscopy to study the structural dynamics of single myosin V molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service