Skip to Content
Merck
All Photos(2)

Key Documents

S3134

Sigma-Aldrich

Streptokinase from β-hemolytic Streptococcus (Lancefield Group C)

lyophilized powder, ≥2,000 units/mg solid

Synonym(s):

Streptokinase from Streptococcus hemolyticus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥2,000 units/mg solid

shipped in

wet ice

storage temp.

−20°C

SMILES string

N1C(CC(C1)C2CCCCC2)C(=O)O

InChI

1S/C11H19NO2/c13-11(14)10-6-9(7-12-10)8-4-2-1-3-5-8/h8-10,12H,1-7H2,(H,13,14)

InChI key

XRZWVSXEDRYQGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell signaling

Streptokinase is an extracellular enzyme produced by β-hemolytic Streptococcus. It is a single-chain polypeptide enzyme that consists of multiple structural domains, including α-, β-, and γ-domains.

Application

Streptokinase has been used in a study to compare primary coronary intervention and thrombolytic therapy in myocardial infarction patients.
Streptokinase is commonly used as a thrombolytic agent in the therapy of ischemic stroke. This therapy carries the important risk of intracerebral hemorrhage. Streptokinase is also used in the treatment of complicated parapneumonic effusions and empyema where adverse reactions, allergic type, are rare.

Biochem/physiol Actions

Streptokinase is a potent plasminogen activator. Unlike human proteins tissue plasminogen activator (tPA) and urokinase plasminogen activator (uPA), microbial streptokinase does not directly activate plasminogen. Instead, it activates plasminogen by fibrin-dependent and fibrin-independent. mechanisms. Streptokinase is effective in thrombolytic therapy. While streptokinase offers promising potential in treating ischemic stroke, its use also presents a significant risk of intracerebral hemorrhage. Streptokinase is also used in treating complicated parapneumonic effusions and empyema where allergic reactions, are rare.

Unit Definition

One unit is equal to the amount of activity in one unit from a Streptokinase International Standard reference vial.

Physical form

Lyophilized powder containing ~50% total protein by biuret and sodium glutamate. Total protein composed of enzyme protein and human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Busch et al.
International journal of molecular sciences, 21(17) (2020-09-06)
During total joint replacement, high concentrations of mesenchymal stromal cells (MSCs) are released at the implantation site. They can be found in cell-tissue composites (CTC) that are regularly removed by surgical suction. A surgical vacuum suction handle was filled with
Role of streptokinase in the treatment of acute loculated parapneumonic pleural effusions and empyema
Bouros D, et al.
Thorax, 49(9), 852-855 (1994)
Ansar S Zhalyalov et al.
PloS one, 12(7), e0180668-e0180668 (2017-07-08)
Fibrinolysis is a cascade of proteolytic reactions occurring in blood and soft tissues, which functions to disintegrate fibrin clots when they are no more needed. In order to elucidate its regulation in space and time, fibrinolysis was investigated using an
Primary coronary intervention versus thrombolytic therapy in myocardial infarction patients in the Middle East
Al-Zakwani, I., et al.
Journal of the American Medical Association, 34(3), 445-451 (2012)
Streptokinase--a clinically useful thrombolytic agent
Banerjee A, et al.
Biotechnology Advances, 22(4), 287-307 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service