Skip to Content
Merck
All Photos(1)

Key Documents

S1763

Sigma-Aldrich

Somatostatin

powder, BioReagent, suitable for cell culture

Synonym(s):

Growth hormone release inhibiting factor, SRIF, Somatostatin-14, Somatotropin release inhibiting factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H104N18O19S2
CAS Number:
Molecular Weight:
1637.88
Beilstein:
6436064
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

product line

BioReagent

Assay

≥97% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]N1[C@@H](Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H]([C@@H](C)O)C(=O)NC(CO)C(=O)N[C@@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](C)N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C1=O)C(O)=O

InChI

1S/C76H104N18O19S2/c1-41(79)64(100)82-37-61(99)83-58-39-114-115-40-59(76(112)113)92-72(108)57(38-95)91-75(111)63(43(3)97)94-71(107)54(33-46-23-11-6-12-24-46)90-74(110)62(42(2)96)93-66(102)51(28-16-18-30-78)84-69(105)55(34-47-36-81-49-26-14-13-25-48(47)49)88-68(104)53(32-45-21-9-5-10-22-45)86-67(103)52(31-44-19-7-4-8-20-44)87-70(106)56(35-60(80)98)89-65(101)50(85-73(58)109)27-15-17-29-77/h4-14,19-26,36,41-43,50-59,62-63,81,95-97H,15-18,27-35,37-40,77-79H2,1-3H3,(H2,80,98)(H,82,100)(H,83,99)(H,84,105)(H,85,109)(H,86,103)(H,87,106)(H,88,104)(H,89,101)(H,90,110)(H,91,111)(H,92,108)(H,93,102)(H,94,107)(H,112,113)/t41-,42+,43+,50-,51-,52-,53-,54-,55-,56-,57?,58-,59-,62-,63-/m0/s1

InChI key

NHXLMOGPVYXJNR-FQSIDJEASA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys

Application

Somatostatin has been used in cell culture. It has also been used in the study of glucose-6-phosphate dehydrogenase activity in monolayer cultures of thyroid epithelial cells.

Biochem/physiol Actions

Somatostatin regulates the endocrine system and affects neurotransmission and cell proliferation via interaction with G protein-coupled somatostatin receptors and inhibition of the release of numerous secondary hormones.
Somatostatin (SST) is a peptide. It performs several functions like anti-proliferation, pro-apoptosis and anti-migration. Somatostatin performs physiological activities with the help of various G protein-coupled receptor subtypes (SST receptor (SSTR)-1, SSTR2A, SSTR2B, and SSTR3–5).

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Reconstitution

To prepare 2 μg/ml stock solution, dissolve in 5 ml sterile culture medium per μg somatostatin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace amine-associated receptor 1 localization at the apical plasma membrane domain of fisher rat thyroid epithelial cells is confined to cilia
Szumska, J, et al.
European thyroid journal, 4(Suppl. 1), 30-41 (2015)
Does somatostatin have a role in the regulation of cortisol secretion in primary pigmented nodular adrenocortical disease (ppnad)? a clinical and in vitro investigation.
Bram Z
The Journal of Clinical Endocrinology and Metabolism, 99(5), E891-E901 (2014)
SSTR2 promoter hypermethylation is associated with the risk and progression of laryngeal squamous cell carcinoma in males
Shen Z, et al.
Diagnostic Pathology, 11(1), 10-10 (2016)
Glucose-6-phosphate dehydrogenase activity in monolayer cultures of thyroid epithelial cells: TSH and inhibition of nitrogen oxide synthase affect the enzyme activity and the oxygen sensitivity of the histochemical assay
Kayser L and Thomsen J
Acta Histochemica, 107(1), 31-41 (2005)
Henrik H Hansen et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 657-664 (2014-07-06)
Type 2 diabetes is characterized by impaired β-cell function associated with progressive reduction of insulin secretion and β-cell mass. Evidently, there is an unmet need for treatments with greater sustainability in β-cell protection and antidiabetic efficacy. Through an insulin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service