Skip to Content
Merck
All Photos(1)

Key Documents

S1195

Sigma-Aldrich

ST638

≥98% (HPLC), solid

Synonym(s):

α-Cyano-(3-ethoxy-4-hydroxy-5-phenylthiomethyl)cinnamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O3S
CAS Number:
Molecular Weight:
354.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

yellow

mp

134-135.5 °C

solubility

DMSO: 19 mg/mL

storage temp.

−20°C

SMILES string

CCOc1cc(cc(CSc2ccccc2)c1O)\C=C(/C#N)C(N)=O

InChI

1S/C19H18N2O3S/c1-2-24-17-10-13(8-14(11-20)19(21)23)9-15(18(17)22)12-25-16-6-4-3-5-7-16/h3-10,22H,2,12H2,1H3,(H2,21,23)/b14-8+

InChI key

YKLMGKWXBLSKPK-RIYZIHGNSA-N

Biochem/physiol Actions

Protein tyrosine kinase inhibitor (IC50 = 370 nM). Also inhibits HGF-induced MAP kinase activation in hepatocytes and inhibits phospholipase D activity in human neutrophils.

Packaging

Light sensitive and packaged under nitrogen.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Green et al.
Biochimica et biophysica acta, 1222(2), 241-248 (1994-06-30)
We have investigated the relationship between tyrosine phosphorylation and respiratory-burst activity in murine bone-marrow-derived macrophages (BMM) stimulated with phorbol myristate acetate (PMA). In unprimed BMM, a good correlation was observed between the net level of tyrosine phosphorylation and the activity
Y Fukumoto et al.
Cardiovascular research, 32(6), 1131-1140 (1996-12-01)
Restenosis after percutaneous transluminal coronary angioplasty (PTCA) still remains a serious late complication. Many growth factors induced in restenotic lesions may be responsible for restenosis after PTCA. Most of the receptors for such growth factors possess tyrosine kinase activity. This
S E Lavens-Phillips et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 47(3), 137-143 (1998-04-30)
To investigate the role of tyrosine kinases (TK) in IgE-mediated signal transduction in human lung mast cells (HLMC) and basophils. Peripheral blood basophils (n > or = 4) and human lung mast cells (n > or = 6). Cells were
R Teshima et al.
Biochimica et biophysica acta, 1221(1), 37-46 (1994-03-10)
We examined the effect of the two protein tyrosine kinase inhibitors, alpha-cyano-3-ethoxy-4-hydroxy-5-phenylthiomethylcinnamide (ST638) and herbimycin A, on the activation processes of rat basophilic leukemia (RBL-2H3) cells by cross-linking of IgE receptors. RBL-2H3 cells sensitized with DNP-specific monoclonal IgE antibody were
A Ito et al.
The Journal of clinical investigation, 96(3), 1288-1294 (1995-09-01)
We recently demonstrated that chronic treatment with IL-1 beta induces coronary arteriosclerotic changes and vasospastic responses to autacoids in pigs in vivo and that those responses are importantly mediated by PDGF. The receptors for PDGF and other major growth factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service