Skip to Content
Merck
All Photos(3)

Key Documents

S8442

Sigma-Aldrich

SU 5416

≥98% (HPLC), powder, neuronal nitric oxide synthase inhibitor

Synonym(s):

1,3-Dihydro-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylene]-2H-indol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O
CAS Number:
Molecular Weight:
238.28
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

SU 5416, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to yellow-orange

mp

221-222 °C

solubility

DMSO: 20 mg/mL, clear (warmed)
H2O: insoluble

storage temp.

−20°C

SMILES string

Cc1cc(C)c(C=C2C(=O)Nc3ccccc23)[nH]1

InChI

1S/C15H14N2O/c1-9-7-10(2)16-14(9)8-12-11-5-3-4-6-13(11)17-15(12)18/h3-8,16H,1-2H3,(H,17,18)/b12-8+

InChI key

WUWDLXZGHZSWQZ-XYOKQWHBSA-N

General description

SU 5416 is a vascular endothelial growth factor (VEGF) receptor protein tyrosine kinase 1/2 inhibitor. SU 5416 is known to prevent tumor growth and tumor angiogenesis.[1] SU 5416 inhibits the activity of neuronal nitric oxide synthase and protects the neuronal cells from nitric oxide-mediated neurotoxicity. It also acts as an agonist of aryl hydrocarbon receptor and may be an effective clinical agent for treating autoimmune diseases and transplant rejection.[2]

Application

SU 5416 has been used to treat HUVEC cells before the tube formation assay to assess its impact on the network generated from tube formation.[3] to induce pulmonary hypertension (PH) in mice to serve as an experimental model[4]

Biochem/physiol Actions

SU 516 inhibits the activity of neuronal nitric oxide synthase and protects the neuronal cells from nitric oxide-mediated neurotoxicity.1 It also acts an agonist of aryl hydrocarbon receptor and is an effective clinical agent for treating autoimmune diseases and transplant rejection.2
SU 5416 is a vascular endothelial growth factor (VEGF) receptor protein tyrosine kinase 1/2 inhibitor.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loredana Ciuclan et al.
American journal of respiratory and critical care medicine, 184(10), 1171-1182 (2011-08-27)
The complex pathologies associated with severe pulmonary arterial hypertension (PAH) in humans have been a challenge to reproduce in mice due to the subtle phenotype displayed to PAH stimuli. Here we aim to develop a novel murine model of PAH
Joshua D Mezrich et al.
PloS one, 7(9), e44547-e44547 (2012-09-13)
The experimental compound SU5416 went as far as Phase III clinical trials as an anticancer agent, putatively because of its activity as a VEGFR-2 inhibitor, but showed poor results. Here, we show that SU5416 is also an aryl hydrocarbon receptor
Aglaia Ntokou et al.
JCI insight, 6(6) (2021-02-17)
Excess macrophages and smooth muscle cells (SMCs) characterize many cardiovascular diseases, but crosstalk between these cell types is poorly defined. Pulmonary hypertension (PH) is a lethal disease in which lung arteriole SMCs proliferate and migrate, coating the normally unmuscularized distal
Florian Ulrich et al.
Developmental biology, 357(1), 134-151 (2011-07-13)
The brain is made of billions of highly metabolically active neurons whose activities provide the seat for cognitive, affective, sensory and motor functions. The cerebral vasculature meets the brain's unusually high demand for oxygen and glucose by providing it with
Charlene Watterston et al.
PLoS genetics, 17(8), e1009769-e1009769 (2021-08-24)
Vessel growth integrates diverse extrinsic signals with intrinsic signaling cascades to coordinate cell migration and sprouting morphogenesis. The pro-angiogenic effects of Vascular Endothelial Growth Factor (VEGF) are carefully controlled during sprouting to generate an efficiently patterned vascular network. We identify

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service