Skip to Content
Merck
All Photos(1)

Documents

S0940

Sigma-Aldrich

β-Cyclodextrin phosphate sodium salt

≥95%

Synonym(s):

Sodium β-cyclodextrin phosphate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

extent of labeling

~4 mol phosphate per mol cyclodextrin

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Used for:
  • Preparation of ionic cluster mimic membranes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service