Skip to Content
Merck
All Photos(1)

Key Documents

R9782

Sigma-Aldrich

RS-1

≥98% (HPLC)

Synonym(s):

3-[(benzylamino)sulfonyl]-4-bromo-N-(4-bromophenyl)benzamide, 4-Bromo-N-(4-bromophenyl)-3-[[(phenylmethyl)amino]sulfonyl]-benzamide, RAD51-stimulatory compound 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Br2N2O3S
CAS Number:
Molecular Weight:
524.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

Brc1ccc(NC(=O)c2ccc(Br)c(c2)S(=O)(=O)NCc3ccccc3)cc1

InChI

1S/C20H16Br2N2O3S/c21-16-7-9-17(10-8-16)24-20(25)15-6-11-18(22)19(12-15)28(26,27)23-13-14-4-2-1-3-5-14/h1-12,23H,13H2,(H,24,25)

InChI key

SWKAVEUTKGKHSR-UHFFFAOYSA-N

Application

RS-1 has been shown to enhance CRISPR genome editing efficiency. To see other small molecule CRISPR enhancers, visit sigma.com/CRISPR-enhancers.
RS-1 has been used:
  • as a homology-directed repair (HDR) agonist to study its effects on in human hematopoietic stem/progenitor cells (HSPCs)
  • as HDR agonist to analyze its effects on DNA repair modulation in human induced pluripotent stem (iPS) cells
  • as RAD51 agonist to study its effects on double-stranded break repair

Biochem/physiol Actions

RS-1 (RAD51-stimulatory compound 1) is a stimulator of the human homologous recombination (HR) protein RAD51. RS-1 stimulates binding of hRAD51 to single stranded DNA (ssDNA) and enhances recombinogenic activity by stabilizing the active form of hRAD51 filaments without inhibiting hRAD51 ATPase activity. RS-1 has been shown to enhance CRISPR-Cas9 knock-in efficiency in HEK293A cells and has been shown to enhance both TALEN and Cas9-mediated knock-in efficiency in rabbits.
RS-1 is a sulfonamido-benzamide compound.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liqian Zhu et al.
Veterinary research, 48(1), 45-45 (2017-09-09)
Bovine herpesvirus 1 (BoHV-1) infection enhanced the generation of inflammatory mediator reactive oxidative species (ROS) and stimulated MAPK signaling that are highly possibly related to virus induced inflammation. In this study, for the first time we show that BoHV-1 infection
Xue Wang et al.
BioMed research international, 2019, 7878906-7878906 (2019-11-07)
It has been reported that paclitaxel administration could cause sensorineural hearing loss, and Wnt activation is important for the development and cell protection of mouse cochlea. However, the effect of Wnt signaling in spiral ganglion neurons (SGNs) damage induced by
Siti Nur Syahirah Ahmad et al.
Foods (Basel, Switzerland), 10(2) (2021-01-31)
This study aims to investigate the effect of different vegetable oils and frying cycles on acrylamide formation during the intermittent frying of beef nuggets. Different vegetable oils, palm olein (PO), red palm olein (RPO), sunflower oil (SFO), and soybean oil
Chieh-Teng Cheng et al.
iScience, 27(4), 109486-109486 (2024-03-29)
Nuclear factor kappa B (NF-κB) is a key regulator in immune signaling and is known to exhibit a digital activation pattern. Yet the molecular basis underlying the heterogeneity in NF-κB activation at single-cell level is not entirely understood. Here, we
Jordan Pinder et al.
Nucleic acids research, 43(19), 9379-9392 (2015-10-03)
CRISPR is a genome-editing platform that makes use of the bacterially-derived endonuclease Cas9 to introduce DNA double-strand breaks at precise locations in the genome using complementary guide RNAs. We developed a nuclear domain knock-in screen, whereby the insertion of a

Articles

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service