Skip to Content
Merck
All Photos(1)

Key Documents

553510

Sigma-Aldrich

RAD51-Stimulatory Compound-1, RS-1

A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis.

Synonym(s):

RAD51-Stimulatory Compound-1, RS-1, 3-((Benzylamino)sulfonyl)-4-bromo-N-(4-bromophenyl)-benzamide, RS-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16Br2N2O3S
CAS Number:
Molecular Weight:
524.23
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

ethanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NCc3ccccc3)c1c(ccc(c1)C(=O)Nc2ccc(cc2)Br)Br

InChI

1S/C20H16Br2N2O3S/c21-16-7-9-17(10-8-16)24-20(25)15-6-11-18(22)19(12-15)28(26,27)23-13-14-4-2-1-3-5-14/h1-12,23H,13H2,(H,24,25)

InChI key

SWKAVEUTKGKHSR-UHFFFAOYSA-N

General description

A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis. Although RS-1 enhances hRAD51 filament formation on ssDNA with or without the cofactor NTP, active filaments and recombinase activity are only induced in the presence of ATP or AMP-PNP, but not with ADP or no cofactors. Shown to promote resistance of primary human neonatal dermal fibroblasts to Cisplatin- (Cat. No. 232120) induced death in a dose-dependent manner. RS-1 is inactive toward related DNA strand exchange proteins scRAD5 and scDMC1 of yeast origin or E. coli RedA.This HDR (homology-directed repair) enhancer, is shown to significantly increase both Cas9 & TALEN-mediated knock-in efficiencies.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up 3 months at -20°C.

Other Notes

Song, J., et al. 2016. Nat. Comm., In Press.
Jayathilaka, K., et al. 2008. Proc. Natl. Acad. Sci. USA.105, 15848.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thao Thu Thi Nguyen et al.
Molecular pharmacology (2023-01-21)
6-mercaptopurine (6-MP) is a key component in maintenance therapy for childhood acute lymphoblastic leukemia (ALL). Recent next-generation sequencing analysis of childhood ALL clarified the emergence of the relapse-specific mutations of the NT5C2 and PRPS1 gene, which are involved in thiopurine
Ya-Wen Fu et al.
Nucleic acids research, 49(2), 969-985 (2021-01-06)
Investigations of CRISPR gene knockout editing profiles have contributed to enhanced precision of editing outcomes. However, for homology-directed repair (HDR) in particular, the editing dynamics and patterns in clinically relevant cells, such as human iPSCs and primary T cells, are
Jacqueline Severino et al.
The EMBO journal, 41(12), e109457-e109457 (2022-05-24)
The mammalian germline is characterized by extensive epigenetic reprogramming during its development into functional eggs and sperm. Specifically, the epigenome requires resetting before parental marks can be established and transmitted to the next generation. In the female germline, X-chromosome inactivation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service