Skip to Content
Merck
All Photos(2)

Key Documents

R5754

Sigma-Aldrich

9-cis-Retinal

vitamin A analog

Synonym(s):

Vitamin A aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.28

Assay

≥95% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

yellow to yellow-orange

mp

56-58 °C (lit.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(=C\C=O)/C=C/C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,15H,7,10,14H2,1-5H3/b9-6+,12-11+,16-8-,17-13+

InChI key

NCYCYZXNIZJOKI-MKOSUFFBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

9-cis-Retinal has been used for visual pigment regeneration.

Biochem/physiol Actions

Retinal, retinol and retinoic acid are the aldehyde, alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain.
9-cis retinal is a natural ligand (chromophore) of the vertebrate rod visual pigment. 9-cis retinal is used in studies on the mechanisms of visual function.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Cambrian origin for vertebrate rods
Sabrina A, et al.
eLife, 4 (2015)
Ching-Ju Tsai et al.
Science advances, 4(9), eaat7052-eaat7052 (2018-09-27)
Selective coupling of G protein (heterotrimeric guanine nucleotide-binding protein)-coupled receptors (GPCRs) to specific Gα-protein subtypes is critical to transform extracellular signals, carried by natural ligands and clinical drugs, into cellular responses. At the center of this transduction event lies the
L Xu et al.
Current opinion in genetics & development, 9(2), 140-147 (1999-05-14)
The nuclear hormone receptors constitute a large family of transcription factors. The binding of the hormonal ligands induces nuclear receptors to assume a configuration that leads to transcriptional activation. Recent studies of retinoic acid and thyroid hormone receptors revealed that
Vladimir J Kefalov et al.
Neuron, 46(6), 879-890 (2005-06-15)
Retinal rod and cone pigments consist of an apoprotein, opsin, covalently linked to a chromophore, 11-cis retinal. Here we demonstrate that the formation of the covalent bond between opsin and 11-cis retinal is reversible in darkness in amphibian red cones
Suddhasil Mookherjee et al.
Cell reports, 25(3), 611-623 (2018-10-18)
Mutations in CEP290 cause ciliogenesis defects, leading to diverse clinical phenotypes, including Leber congenital amaurosis (LCA). Gene therapy for CEP290-associated diseases is hindered by the 7.4 kb CEP290 coding sequence, which is difficult to deliver in vivo. The multi-domain structure of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service