Skip to Content
Merck
All Photos(3)

Documents

B1381

Sigma-Aldrich

8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt

≥98% (HPLC), powder

Synonym(s):

8-Br-cGMP, 8-Bromo-cyclic GMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrN5NaO7P
CAS Number:
Molecular Weight:
446.08
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na+].NC1=Nc2c(nc(Br)n2[C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C(=O)N1

InChI

1S/C10H11BrN5O7P.Na/c11-9-13-3-6(14-10(12)15-7(3)18)16(9)8-4(17)5-2(22-8)1-21-24(19,20)23-5;/h2,4-5,8,17H,1H2,(H,19,20)(H3,12,14,15,18);/q;+1/p-1/t2-,4-,5-,8-;/m1./s1

InChI key

ZJRFCXHKYQVNFK-YEOHUATISA-M

General description

8-Bromoguanosine 3′,5′-cyclic monophosphate is a cell-permeable cGMP analog of cyclic guanosine 3′:5′-monophosphate (cGMP). It is a lipid-soluble analog used in cGMP based contraction studies in cardiomyocytes.

Application

8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt has been used:
  • as a cyclic guanosine 3′:5′-monophosphate (cGMP) agonist in the collapse assay in retinal ganglion axons(10)
  • as a component of reaction buffer in in vitro kinase activity assay of recombinant protein kinase (PKG)(11)
  • as a cyclic nucleotide analog for the induction of cyclic nucleotide-gated channel (CNGA and CNGC) expression in human embryonic kidney cells(12)

Biochem/physiol Actions

8-Bromoguanosine 3′,5′-cyclic monophosphate has greater resistance to hydrolysis by phosphodiesterases than cGMP. Activates cGMP-dependent protein kinase. It slows or inhibits the intracellular calcium oscillations of tracheal smooth muscle cells in response to acetylcholine. 8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt mimics the effect of nitric oxide generating drugs. It modulates circadian rhythms and increases optic nerve impulses. In the eye 8-Bromoguanosine 3′,5′-cyclic monophosphate may favour transduction of non-R-type photoreceptors.
Cell-permeable cGMP analog having greater resistance to hydrolysis by phosphodiesterases than cGMP. Activates cGMP-dependent protein kinase. Slows or inhibits the intracellular calcium oscillations of tracheal smooth muscle cells in response to acetylcholine. Reported to mimic the effect of nitric oxide generating drugs.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U C Garg et al.
The Journal of clinical investigation, 83(5), 1774-1777 (1989-05-01)
Endothelium-derived relaxing factor has been recently identified as nitric oxide. The purpose of this study was to determine if vasodilator drugs that generate nitric oxide inhibit vascular smooth muscle mitogenesis and proliferation in culture. Three chemically dissimilar vasodilators, sodium nitroprusside
8-bromo-cGMP reduces the myofilament response to Ca2+ in intact cardiac myocytes.
Shah AM, et al.
Circulation Research, 74(5), 970-978 (1994)
Cyclic guanosine 3': 5'-monophosphate mimics the effects of light on a circadian pacemaker in the eye of aplysia
Eskin A, et al.
The Journal of Neuroscience, 4(10), 2466-2471 (1984)
Nadine Suffee et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(5), E771-E780 (2017-01-18)
The abundance of epicardial adipose tissue (EAT) is associated with atrial fibrillation (AF), the most frequent cardiac arrhythmia. However, both the origin and the factors involved in EAT expansion are unknown. Here, we found that adult human atrial epicardial cells
Karla Kretschmannova et al.
Molecular endocrinology (Baltimore, Md.), 26(1), 153-164 (2011-12-03)
Pituitary cells fire action potentials independently of external stimuli, and such spontaneous electrical activity is modulated by a large variety of hypothalamic and intrapituitary agonists. Here, we focused on the potential role of hyperpolarization-activated and cyclic nucleotide-gated (HCN) channels in

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service