Skip to Content
Merck
All Photos(2)

Key Documents

R4528

Sigma-Aldrich

Anti-hnRNP-A1 antibody, Mouse monoclonal

clone 9H10, purified from hybridoma cell culture

Synonym(s):

Anti-heterogeneous nuclear ribonucleoprotein-A1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9H10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~34 kDa

species reactivity

human, hamster, monkey, rat, bovine, mouse, canine

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 0.5-1.0 μg/mL using total cell extract of 293T cells

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

hnRNP-A1 (heterogeneous nuclear ribonucleoprotein) is a pre-mRNA/mRNA binding protein, that belongs to the hnRNP family. It is located in chromosome 12q13.1. It is located in the nucleoplasm but it travels between the nucleus and the cytoplasm. It has two RNP motif RNA-binding domains (RBDs) at the amino terminus and a glycine-rich domain at the carboxyl terminus.

Immunogen

purified human hnRNP-A1.

Application

Anti-hnRNP-A1 has been used in immunofluorescence, immunohistochemistry and western blotting.

Does not immunoprecipitate the hnRNP complex.

Biochem/physiol Actions

hnRNP-A1 (heterogeneous nuclear ribonucleoprotein) participates in several RNA metabolisms. It also participates in deep vein thrombosis patients with behçet′s disease. hnRNPA1 mutations results in amyotrophic lateral sclerosis and multisystem proteinopathy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HnRNP A1 is Involved in Deep Vein Thrombosis Patients with Behcet's Disease
Chen P, et al.
EBioMedicine, 6, 215-221 (2016)
Carolina Villarroya-Beltri et al.
Nature communications, 4, 2980-2980 (2013-12-21)
Exosomes are released by most cells to the extracellular environment and are involved in cell-to-cell communication. Exosomes contain specific repertoires of mRNAs, microRNAs (miRNAs) and other non-coding RNAs that can be functionally transferred to recipient cells. However, the mechanisms that
Gabriel S Lopes et al.
Molecular biology reports, 49(6), 4257-4268 (2022-02-23)
We have identified endogenous p65 to be an SDS-stable dimer protein composed of ~ 37 kDa hnRNPA/B-like subunits. We have investigated molecular properties involved in the stability of dimeric form, and their regulation in the transition between monomeric and dimeric forms of hnRNPA/B-like
Nagalakshmi Nadiminty et al.
Molecular cancer therapeutics, 14(8), 1884-1895 (2015-06-10)
Castration-resistant prostate cancer (CRPC) remains dependent on androgen receptor (AR) signaling. Alternative splicing of the AR to generate constitutively active, ligand-independent variants is one of the principal mechanisms that promote the development of resistance to next-generation antiandrogens such as enzalutamide.
Simon Braun et al.
Nature communications, 9(1), 3315-3315 (2018-08-19)
Mutations causing aberrant splicing are frequently implicated in human diseases including cancer. Here, we establish a high-throughput screen of randomly mutated minigenes to decode the cis-regulatory landscape that determines alternative splicing of exon 11 in the proto-oncogene MST1R (RON). Mathematical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service