Skip to Content
Merck
All Photos(1)

Key Documents

05-1521

Sigma-Aldrich

Anti-hnRNP A1 Antibody, clone 4B10

clone 4B10, from mouse

Synonym(s):

Helix-destabilizing protein, Single-strand RNA-binding protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B10, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HNRNPA1(3178)

General description

The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA (hnRNA) and associate with pre-mRNAs in the nucleus. These complexes are associated with pre-mRNA processing and other aspects of mRNA metabolism and transport. While all hnRNPs are present in the nucleus, data suggests they shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by hnRNP A1 has two repeats of quasi-RRM domains that bind to RNAs. This abundant core protein, along with other hnRNP proteins, is exported from the nucleus, probably bound to mRNA, and is immediately re-imported. The hnRNP A1 protein is involved in the packaging of pre-mRNA into hnRNP particles, transport of poly A+ mRNA from the nucleus to the cytoplasm, and may have a role in splice site selection.

Specificity

This antibody recognizes hnRNP A1.

Immunogen

Epitope: Unknown
Full length native protein partially purified human hnRNP A1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-hnRNP A1 Antibody, clone 4B10 (mouse monoclonal antibody) validated in WB to detect hnRNP A1 also known as Helix-destabilizing protein, Single-str& RNA-binding protein, heterogeneous nuclear ribonucleoprotein A1.

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.1 µg/ml of this antibody detected HnRNP A1 on 10 µg of K562 cell lysate.

Target description

~ 38 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah E Salapa et al.
Journal of neuroscience research, 98(4), 704-717 (2019-11-23)
Altered stress granule (SG) and RNA-binding protein (RBP) biology have been shown to contribute to the pathogenesis of several neurodegenerative diseases, yet little is known about their role in multiple sclerosis (MS). Pathological features associated with dysfunctional RBPs include RBP
Joseph P Clarke et al.
Frontiers in molecular biosciences, 10, 1178439-1178439 (2023-07-10)
The RNA binding protein heterogeneous nuclear ribonucleoprotein A1 (A1) regulates RNA metabolism, which is crucial to maintaining cellular homeostasis. A1 dysfunction mechanistically contributes to reduced cell viability and loss, but molecular mechanisms of how A1 dysfunction affects cell viability and
Joseph-Patrick W E Clarke et al.
International journal of molecular sciences, 22(6) (2021-04-04)
Evidence indicates that dysfunctional heterogeneous ribonucleoprotein A1 (hnRNPA1; A1) contributes to the pathogenesis of neurodegeneration in multiple sclerosis. Understanding molecular mechanisms of neurodegeneration in multiple sclerosis may result in novel therapies that attenuate neurodegeneration, thereby improving the lives of MS
Dooyoung Lee et al.
RNA (New York, N.Y.), 23(7), 1035-1047 (2017-04-13)
The nuclear RNase III enzyme DROSHA interacts with its cofactor DGCR8 to form the Microprocessor complex, which initiates microRNA (miRNA) maturation by cleaving hairpin structures embedded in primary transcripts. Apart from its central role in the biogenesis of miRNAs, DROSHA
Joshua N Douglas et al.
Journal of clinical & cellular immunology, 7(2), 402-402 (2016-07-05)
Multiple sclerosis (MS) is the most common demyelinating disorder of the central nervous system (CNS). Data suggest that antibodies to CNS targets contribute to the pathogenesis of MS. MS patients produce autoantibodies to heterogeneous nuclear ribonucleoprotein A1 (hnRNP A1). hnRNP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service