Skip to Content
Merck
All Photos(1)

Key Documents

444810

Sigma-Aldrich

ProteoExtract® Native Membrane Protein Extraction Kit

Synonym(s):

Membrane protein extraction kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.77

usage

sufficient for 20 extractions

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

protein extraction: suitable

input

sample type: mammalian tissue

shipped in

ambient

storage temp.

2-8°C

General description

A convenient kit for the isolation of native integral membrane and membrane-associated proteins, without the need for ultracentrifugation. Extraction is based on association of proteins with cellular membranes rather than on their hydrophobicity. Resulting samples are suitable for use in functional assays, 2-D gel electrophoresis, and other applications.
Membrane proteins represent only one-third of the proteins encoded by the human genome, but they represent more than two-thirds of the known protein targets for drugs. Therefore, approaches to prepare and characterize membrane proteins are of significant interest for drug discovery. However, due to their hydrophobic nature, membrane proteins are more difficult to analyze than soluble proteins. In addition to the intrinsic difficulty of solubilization, membrane proteins are challenging because of their general low abundance. Thus, the challenges for sample preparation of membrane proteins are effective solubilization and selective enrichment, ideally keeping proteins in a non-denatured state.
Note: 1 kit is sufficient for up to 20 sample extractions.

Components

Wash Buffer, Extraction Buffer 1, Extraction Buffer 2, Protease Inhibitor Cocktail, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Principle

The ProteoExtract® Native Membrane Protein Extraction Kit is designed for the isolation of native membrane proteins from a broad range of mammalian samples, including:

• Adherent tissue culture cells

• Suspension grown tissue culture cells

• Frozen cell pellets

• Tissues

Preparation Note

The amount of buffer required for each extraction is dependent upon the amount of starting cell material.

Storage and Stability

• Wash Buffer

Store at 4°C.

• Extraction Buffers I & II

The Extraction Buffers I and II can be stored at 4°C for up to 6 month.

For prolonged storage, freeze the buffers in convenient aliquots at -20°C. Before extraction, buffers must be thawed at room temperature (RT). After thawing, mix components by gently shaking or vortexing. Avoid repeated freezing and thawing!

• Protease Inhibitor Cocktail

The Protease Inhibitor Cocktail is supplied in DMSO and can be stored at 4°C up to 6 months. For prolonged storage, freeze the cocktail in convenient aliquots at -20°C. During the sample preparation procedure it must be kept at RT to prevent freezing of DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robyn T Sussman et al.
Frontiers in oncology, 10, 302-302 (2020-03-27)
We developed a computational pipeline designed to use RNA sequencing (n = 136) and gene expression profiling (n = 250) data from neuroblastoma tumors to identify cell surface proteins predicted to be highly expressed in MYCN amplified neuroblastomas and with
Meenakshi Upreti et al.
Journal of molecular medicine (Berlin, Germany), 91(4), 497-506 (2012-10-24)
The present study reports on a new strategy for selective, radiation therapy-amplified drug delivery using an antiangiogenic 33-a.a., tumor vasculature-targeting ligand, anginex, to improve the therapeutic ratio for strategies developed against solid tumors. Our findings indicate that galectin-1 is (a)
Jacqueline Bezençon et al.
Journal of pharmaceutical sciences, 110(1), 404-411 (2020-10-16)
Recent studies have focused on coproporphyrin (CP)-I and CP-III (CPs) as endogenous biomarkers for organic anion transporting polypeptides (OATPs). Previous data showed that CPs are also substrates of multidrug resistance-associated protein (MRP/Mrp) 2 and 3. This study was designed to
Wenlu Li et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 39(10), 2048-2060 (2018-05-23)
Blood-brain barrier (BBB) disruption in neurological disorders remains an intractable problem with limited therapeutic options. Here, we investigate whether the endothelial cell membrane protein annexin A2 (ANXA2) may play a role in reducing trans-endothelial permeability and maintaining cerebrovascular integrity after
Raju Khatri et al.
Journal of pharmacological and toxicological methods, 98, 106590-106590 (2019-06-04)
Sandwich-cultured human hepatocytes (SCHHs) are the most common in vitro hepatocyte model used for studying hepatic drug disposition and hepatotoxicity. Targeted quantification of key DME and transporter protein expression is useful for in vitro-in vivo extrapolation of drug and xenobiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service