Skip to Content
Merck
All Photos(1)

Key Documents

PZ0190

Sigma-Aldrich

Avasimibe

≥98% (HPLC)

Synonym(s):

CI-1011; PD 148515; [[2,4,6-tris(1-methylethyl)phenyl]acetyl]-, 2,6-bis(1-methylethyl)phenyl ester] sulfamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H43NO4S
CAS Number:
Molecular Weight:
501.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥40 mg/mL

relevant disease(s)

Alzheimer′s disease; cardiovascular diseases

storage temp.

room temp

SMILES string

CC(C)c1cc(C(C)C)c(CC(=O)NS(=O)(=O)Oc2c(cccc2C(C)C)C(C)C)c(c1)C(C)C

InChI

1S/C29H43NO4S/c1-17(2)22-14-25(20(7)8)27(26(15-22)21(9)10)16-28(31)30-35(32,33)34-29-23(18(3)4)12-11-13-24(29)19(5)6/h11-15,17-21H,16H2,1-10H3,(H,30,31)

InChI key

PTQXTEKSNBVPQJ-UHFFFAOYSA-N

Related Categories

Application

Avasimibe has been used as an inhibitor of acyl-coenzyme A: cholesterol acyltransferase (ACAT) in Huh7.5.1 cells for testing its combination with direct-acting antivirals (DAAs) and to test its effect on lipid droplet accumulation in acidosis-adapted cancer cells. It may be used as an inhibitor of ACAT to assess cholesterol esterification in Trypanosoma cruzi.

Biochem/physiol Actions

Avasimibe (CI-1011) is an orally bioavailable Acyl-CoA:Cholesterol O-Acyltransferase (ACAT) inhibitor. It was originally developed as an antilepic drug, and was shown to significantly reduce plasma total triglyceride and VLDL-cholesterol, but later clinical trials were disappointing. ACAT has also been investigated as a potential therapeutic target for Alzheimer′s disease. Recent studies have looked at the effects of avasimibe in reducing amyloid pathology by limiting generation and increasing clearance of diffusible amyloid-beta (Abeta).
Avasimibe is an orally bioavailable Acyl-CoA:Cholesterol O-Acyltransferase (ACAT) inhibitor

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasminder Sahi et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(12), 1370-1376 (2004-08-31)
Avasimibe, an acyl-CoA:cholesterol acyltransferase inhibitor, has been previously shown to be a potent inducer of CYP3A4 and multiple drug resistance protein 1. We have further characterized the drug interaction potential of avasimibe by studying the inductive and inhibitory effect of
Frederick J Raal et al.
Atherosclerosis, 171(2), 273-279 (2003-12-04)
This study assessed the efficacy and safety of avasimibe (CI-1011), an inhibitor of acyl coenzyme A-cholesterol acyltransferase (ACAT) in subjects with homozygous familial hypercholesterolemia (HoFH). Twenty seven subjects were enrolled in a double-blind, randomized, 3-sequence crossover trial of atorvastatin 80
Jean-Claude Tardif et al.
The American journal of cardiology, 98(1), 23-27 (2006-06-21)
We assessed vascular changes during atherosclerosis regression. Compensatory enlargement of coronary arteries accommodates plaque burden during atherosclerosis development. Lipid-lowering therapy has altered the natural history of coronary atherosclerosis, but the arterial changes that occur during disease regression need to be
Jean-Claude Tardif et al.
Circulation, 110(21), 3372-3377 (2004-11-10)
Inhibition of the acyl coenzyme A:cholesterol acyltransferase (ACAT) enzyme may prevent excess accumulation of cholesteryl esters in macrophages. The ACAT inhibitor avasimibe was shown to reduce experimental atherosclerosis. This study was designed to investigate the effects of avasimibe on human
Yves Rival et al.
DNA and cell biology, 23(5), 283-292 (2004-06-01)
It is now recognized that atherosclerosis complications are related to the unstable character of the plaque rather than its volume. Vulnerable plaques often contain a large lipid core, a reduced content of smooth muscle cells, and accumulation of inflammatory cells.

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service