Skip to Content
Merck
All Photos(1)

Key Documents

SML1648

Sigma-Aldrich

24(S)-Hydroxycholesterol

≥98% (HPLC)

Synonym(s):

24S-Hydroxycholesterol, 5-Cholesten-3β,24(S)-diol, Cerebrosterin, Cerebrosterol, Cholest-5-ene-3β,24-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -45 to -52°, c = 1 in chloroform-d

storage condition

desiccated

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CC[C@H](O)C(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25+,26+,27-/m1/s1

InChI key

IOWMKBFJCNLRTC-XWXSNNQWSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

24(S)-Hydroxycholesterol is the major brain metabolite formed from cholesterol in the brain and is important for brain cholesterol homeostasis in addition to being an an endogenous agonist of the nuclear liver X receptors (LXRs).
24(S)-Hydroxycholesterol is the major brain metabolite formed from cholesterol in the brain and is important for brain cholesterol homeostasis in addition to being an an endogenous agonist of the nuclear liver X receptors (LXRs). Impairment of brain cholesterol metabolism has been associated with several neurodegenerative diseases, including multiple sclerosis, Alzheimer′s, and Huntington diseases. The exact role role of 24(S)-Hydroxycholesterol is unclear, and it may have differing effects depending on concentration. Low concentrations can protect neuronal cells against oxidative stress, while high concentrations induce necroptosis-like neuronal cell death. 24(S)-Hydroxycholesterol has also been found to be a potent allosteric modulator of N-methyl-d-aspartate receptors, potentiating NMDAR-mediated EPSCs in rat hippocampal neurons with an EC50 of 1.2 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elza Fonseca et al.
Genome biology and evolution, 9(1), 222-230 (2017-01-07)
Nuclear receptors (NRs) regulate numerous aspects of the endocrine system. They mediate endogenous and exogenous cues, ensuring a homeostatic control of development and metabolism. Gene duplication, loss and mutation have shaped the repertoire and function of NRs in metazoans. Here
Genta Kakiyama et al.
The Journal of steroid biochemistry and molecular biology, 189, 36-47 (2019-02-03)
The aim of this paper was to more completely study the mitochondrial CYP27A1 initiated acidic pathway of cholesterol metabolism. The mitochondrial CYP27A1 initiated pathway of cholesterol metabolism (acidic pathway) is known to synthesize two well-described vital regulators of cholesterol/lipid homeostasis
Steven M Paul et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(44), 17290-17300 (2013-11-01)
N-methyl-D-aspartate receptors (NMDARs) are glutamate-gated ion channels that are critical to the regulation of excitatory synaptic function in the CNS. NMDARs govern experience-dependent synaptic plasticity and have been implicated in the pathophysiology of various neuropsychiatric disorders including the cognitive deficits
Paola Gamba et al.
Aging cell, 13(3), 561-572 (2014-03-13)
An abnormal accumulation of cholesterol oxidation products in the brain of patients with Alzheimer's disease (AD) would further link an impaired cholesterol metabolism in the pathogenesis of the disease. The first evidence stemming from the content of oxysterols in autopsy
Xiaofei Wei et al.
Neuropharmacology, 148, 11-20 (2018-12-31)
24S-hydroxycholesterol (24HC) is the major metabolic breakdown product of cholesterol in the brain. Among its other effects on neurons, 24HC modulates N-methyl-d-aspartate (NMDA or GluN) receptors, but our understanding of this mechanism is poor. We used whole-cell patch clamp recordings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service