Skip to Content
Merck
All Photos(1)

Documents

PZ0020

Sigma-Aldrich

Temsirolimus

≥98% (HPLC)

Synonym(s):

42-[3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoate]-rapamycin, CCI-779

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H87NO16
CAS Number:
Molecular Weight:
1030.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CO[C@@H]1CC(CC[C@H]1OC(=O)C(C)(CO)CO)C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(C)\[C@H](CC3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCCC4C(=O)O2)OC

InChI

1S/C56H87NO16/c1-33-17-13-12-14-18-34(2)45(68-9)29-41-22-20-39(7)56(67,73-41)51(63)52(64)57-24-16-15-19-42(57)53(65)71-46(30-43(60)35(3)26-38(6)49(62)50(70-11)48(61)37(5)25-33)36(4)27-40-21-23-44(47(28-40)69-10)72-54(66)55(8,31-58)32-59/h12-14,17-18,26,33,35-37,39-42,44-47,49-50,58-59,62,67H,15-16,19-25,27-32H2,1-11H3/b14-12+,17-13+,34-18+,38-26+/t33-,35-,36-,37-,39-,40+,41+,42+,44-,45+,46+,47-,49-,50+,56-/m1/s1

InChI key

CBPNZQVSJQDFBE-FUXHJELOSA-N

Gene Information

human ... FKBP1A(2280)

General description

Temsirolimus is an esterified rapamycin compound, soluble in water.

Application

Temsirolimus has been used:
  • in combination with radiation to study its effect on von Hippel Lindau (VHL) tumour suppressor gene-deficient renal carcinoma cell lines
  • to induce autophagy
  • as a mammalian target of rapamycin complex 1 (mTORC1) inhibitor to study the effect of cellular Akt-kinase on human papillomaviruses 16 (HPV16) late gene expression

Biochem/physiol Actions

Temsirolimus is a specific inhibitor of mammalian target of rapamycin (mTOR) mTOR Complex 1 (mTORC1). Temsirolimus is an antiproliferative and antiangiogenic, the first-in-class mTOR inhibitor approved for the treatment of patients with advanced poor prognosis renal cell carcinoma.
The inhibitory action of temsirolimus is mediated by FKBP-12 (FK506-binding protein 12), a major intracellular protein. It is useful in treating renal-cell carcinoma.

Features and Benefits

This compound is featured on the PKB/Akt page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Guilbert et al.
PloS one, 8(12), e85995-e85995 (2014-01-07)
Inhibitors of the mammalian target of rapamycin (mTORi) have clinical activity; however, the benefits of mTOR inhibition by rapamycin and rapamycin-derivatives (rapalogs) may be limited by a feedback mechanism that results in AKT activation. Increased AKT activity resulting from mTOR
Temsirolimus, interferon alfa, or both for advanced renal-cell carcinoma
Hudes G, et al.
The New England Journal of Medicine, 356(22), 2271-2281 (2007)
Viktor Grünwald et al.
European urology, 70(6), 1006-1015 (2016-05-31)
The predictive role of objective remission remains undefined for targeted agents in metastatic renal cell carcinoma (mRCC); however, early tumour shrinkage (eTS) was shown to be predictive and/or prognostic for overall survival (OS) and progression-free survival (PFS) in mRCC in
Temsirolimus
Ma WW and Jimeno A
Drugs Today (Barc), 43(10), 659-669 (2007)
C Le Tourneau et al.
British journal of cancer, 99(8), 1197-1203 (2008-09-18)
The proof of principle that a drug targeting mTOR can improve survival has been obtained recently from a large randomised trial using temsirolimus as a first-line therapy in patients with advanced poor prognostic renal cell carcinoma. Consistent data have recently

Articles

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

We offer many products related to PKB/Akt for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service