Skip to Content
Merck
All Photos(2)

Key Documents

PLA0057

Sigma-Aldrich

Rabbit anti-Mre11 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

AT-like disease, ATLD, DNA recombination and repair protein, HNGS1, MRE11, MRE11 double strand break repair nuclease A, MRE11 homolog, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11 meiotic recombination 11-like protein A, MRE11B, double strand break repair nuclease A, endo/exonuclease Mre11, meiotic recombination (S. cerevisiae) 11 homolog A, meiotic recombination 11 homolog 1, meiotic recombination 11 homolog A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:1,000-1:5,000
western blot: 1:1,000- 1:5,000

accession no.

NP_005582.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Mre11(4361)

Immunogen

The epitope recognized by PLA0057 maps to a region between residues 1 and 50 of human Meiotic Recombination 11 homolog A using the numbering given in entry NP_005582.1 (GeneID 4361).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Mre11 is a component of the MRN complex and plays a role in telomere maintenance and double-strand break (DSB) repair. Mre11 associates with rad50 in the MRN complex. Mre11 possesses 3′– 5′ exonuclease activity that is involved in creating DNA termini in DSB suitable for priming DNA synthesis and ligation.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beeke Wienert et al.
Science (New York, N.Y.), 364(6437), 286-289 (2019-04-20)
CRISPR-Cas genome editing induces targeted DNA damage but can also affect off-target sites. Current off-target discovery methods work using purified DNA or specific cellular models but are incapable of direct detection in vivo. We developed DISCOVER-Seq (discovery of in situ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service