Skip to Content
Merck
All Photos(2)

Documents

PEPTIDE

Custom Peptides

Product Overview, Design, and Ordering

Synonym(s):

AQUA peptide, Isotopically labeled peptide, PEPscreen, peptide array, peptide library, peptide microarray

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105906
Magdalena Banas et al.
PloS one, 8(3), e58709-e58709 (2013-03-26)
Chemerin, a chemoattractant ligand for chemokine-like receptor 1 (CMKLR1) is predicted to share similar tertiary structure with antibacterial cathelicidins. Recombinant chemerin has antimicrobial activity. Here we show that endogenous chemerin is abundant in human epidermis, and that inhibition of bacteria
Joyce J C Kroot et al.
American journal of hematology, 87(10), 977-983 (2012-08-14)
Measurements of the iron regulatory hormone hepcidin by various methodologies and laboratories are not harmonized. As a result different numeric results are obtained for the same clinical sample. We investigated whether better agreement between plasma hepcidin methods can be achieved
Linn S Strandberg et al.
PloS one, 8(9), e72668-e72668 (2013-09-17)
Congenital heart block (CHB) is a transplacentally acquired autoimmune disease associated with anti-Ro/SSA and anti-La/SSB maternal autoantibodies and is characterized primarily by atrioventricular (AV) block of the fetal heart. This study aims to investigate whether the T-type calcium channel subunit
Artur Schmidtchen et al.
The Journal of biological chemistry, 284(26), 17584-17594 (2009-04-29)
A novel approach for boosting antimicrobial peptides through end tagging with hydrophobic oligopeptide stretches is demonstrated. Focusing on two peptides derived from kininogen, GKHKNKGKKNGKHNGWK (GKH17) and HKHGHGHGKHKNKGKKN (HKH17), tagging resulted in enhanced killing of Gram-positive Staphylococcus aureus, Gram-negative Escherichia coli
Arminja N Kettenbach et al.
Nature protocols, 6(2), 175-186 (2011-02-05)
In the analysis of biological systems, it is of interest to identify the components of the system and to monitor their changes in abundance under different conditions. The AQUA (for 'absolute quantification') method allows sensitive and specific targeted quantification of

Articles

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Consider sequence length, solubility, and stability when designing peptides for custom synthesis.

Related Content

Peptide synthesis up to 200 amino acids with HPLC purification, offering various purity levels, scales, and modifications.

Peptide synthesis up to 200 amino acids with HPLC purification, offering various purity levels, scales, and modifications.

Peptide synthesis up to 200 amino acids with HPLC purification, offering various purity levels, scales, and modifications.

Peptide synthesis up to 200 amino acids with HPLC purification, offering various purity levels, scales, and modifications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service