Skip to Content
Merck
All Photos(1)

Key Documents

T3410

Sigma-Aldrich

Thymosin α1 bovine

≥90% (HPLC)

Synonym(s):

Ac-Ser-Asp-Ala-Ala-Val-Asp-Thr-Ser-Ser-Glu-Ile-Thr-Thr-Lys-Asp-Leu-Lys-Glu-Lys-Lys-Glu-Val-Val-Glu-Glu-Ala-Glu-Asn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C129H215N33O55
CAS Number:
Molecular Weight:
3108.28
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥90% (HPLC)

form

powder

color

white

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CO)NC(C)=O)C(C)C)[C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C129H215N33O55/c1-18-59(10)98(159-114(201)76(36-42-91(181)182)146-120(207)83(53-164)154-121(208)84(54-165)155-127(214)99(63(14)166)160-118(205)80(51-94(187)188)152-123(210)95(56(4)5)156-104(191)62(13)135-102(189)60(11)137-115(202)78(49-92(183)184)151-119(206)82(52-163)138-66(17)169)125(212)161-101(65(16)168)128(215)162-100(64(15)167)126(213)147-70(30-22-26-46-133)109(196)150-79(50-93(185)186)117(204)149-77(47-55(2)3)116(203)142-69(29-21-25-45-132)108(195)144-73(33-39-88(175)176)110(197)141-67(27-19-23-43-130)106(193)140-68(28-20-24-44-131)107(194)145-75(35-41-90(179)180)113(200)157-97(58(8)9)124(211)158-96(57(6)7)122(209)148-74(34-40-89(177)178)111(198)143-71(31-37-86(171)172)105(192)136-61(12)103(190)139-72(32-38-87(173)174)112(199)153-81(129(216)217)48-85(134)170/h55-65,67-84,95-101,163-168H,18-54,130-133H2,1-17H3,(H2,134,170)(H,135,189)(H,136,192)(H,137,202)(H,138,169)(H,139,190)(H,140,193)(H,141,197)(H,142,203)(H,143,198)(H,144,195)(H,145,194)(H,146,207)(H,147,213)(H,148,209)(H,149,204)(H,150,196)(H,151,206)(H,152,210)(H,153,199)(H,154,208)(H,155,214)(H,156,191)(H,157,200)(H,158,211)(H,159,201)(H,160,205)(H,161,212)(H,162,215)(H,171,172)(H,173,174)(H,175,176)(H,177,178)(H,179,180)(H,181,182)(H,183,184)(H,185,186)(H,187,188)(H,216,217)/t59-,60-,61-,62-,63+,64+,65+,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,95-,96-,97-,98-,99-,100-,101-/m0/s1

InChI key

NZVYCXVTEHPMHE-ZSUJOUNUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hormone produced by thymic stromal cells that can augment host defense mechanisms, by increasing the production of T cells by thymus and restoring T cell-mediated antibody production in immunodeficient animals..

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Ying Zhang et al.
Protein expression and purification, 77(2), 140-145 (2011-01-25)
In this study, a human thymosin-α1 (hTα1) fusion protein was overexpressed in Escherichia coli (E. coli). The hexahistidine-tagged hTα1 fusion protein was obtained in soluble form in cells of the engineered E. coli strain BL21 (DE3)/pET-28a-hTα1 that had been induced
Concepción S Sarandeses et al.
The Journal of biological chemistry, 278(15), 13286-13293 (2003-01-30)
Thymosin alpha(1) (T alpha(1)) and thymosin T alpha(11) (T alpha(11)) are polypeptides with immunoregulatory properties first isolated from thymic extracts, corresponding to the first 28 and 35 amino acid residues, respectively, of prothymosin alpha (ProT alpha), a protein involved in
Longxin Qiu et al.
International journal of molecular medicine, 23(5), 597-602 (2009-04-11)
We investigated the effects of the in vivo administration of thymosin alpha-1 (Talpha-1) on streptozotocin (STZ)-induced pancreatic lesions and diabetes. Mice were randomly divided into four experimental groups: normoglycemic control, STZ-treated, STZ plus 0.1 microg/kg body weight/day Talpha-1-treated, and STZ
Juan Li et al.
Peptides, 31(11), 2151-2158 (2010-08-12)
Thymosin alpha 1 (Tα1), a 28-amino acid peptide, was first described and characterized from calf thymuses in 1977. This peptide can enhance T-cell, dendritic cell (DC) and antibody responses, modulate cytokines and chemokines production and block steroid-induced apoptosis of thymocytes.
Luigina Romani et al.
Annals of the New York Academy of Sciences, 1112, 326-338 (2007-05-15)
Thymosin alpha1 (Talpha1), first described and characterized by Allan Goldstein in 1972, is used worldwide for the treatment of some immunodeficiencies, malignancies, and infections. Although Talpha1 has shown a variety of effects on cells and pathways of the immune system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service