Skip to Content
Merck
All Photos(1)

Documents

P9752

Sigma-Aldrich

Phosphoriboisomerase from spinach

Type I, partially purified powder, ≥40 units/mg protein (biuret)

Synonym(s):

D-Ribose-5-phosphate ketol-isomerase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

spinach

Quality Level

type

Type I

form

partially purified powder

specific activity

≥40 units/mg protein (biuret)

storage temp.

−20°C

Application

Phosphoriboisomerase is used to study the ribulose monophosphate pathway. Phosphoriboisomerase from spinach is used in enzyme assays to convert ribose 5-phosphate to ribulose 5-phosphate.

Biochem/physiol Actions

Phosphoriboisomerase is responsible for the converstion of ribose 5-phosphate to ribulose 5-phosphate.

Unit Definition

One unit will convert 1.0 μmole of D-ribose 5-phosphate to D-ribulose 5-phosphate per min at pH 7.7 at 30 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Soo-Jin Yeom et al.
Biotechnology letters, 32(6), 829-835 (2010-02-16)
The activity of ribose-5-phosphate isomerases (RpiB) from Clostridium difficile for D-ribose isomerization was optimal at pH 7.5 and 40 degrees C, while that from Thermotoga maritima for L-talose isomerization was optimal at pH 8.0 and 70 degrees C. C. difficile
Anna Tylki-Szymańska et al.
Postepy biochemii, 57(2), 168-171 (2011-09-15)
Two previously unreported inborn errors of metabolism occur in the reversible part of the pentose phosphate pathway. Deficiency of ribose-5-phosphate isomerase has been described in one patient who suffered from a progressive leukoencephalopathy and developmental delay. Transaldolase deficiency has been
Sandrine Mariano et al.
Carbohydrate research, 344(7), 869-880 (2009-03-31)
This study reports syntheses of d-allose 6-phosphate (All6P), D-allulose (or D-psicose) 6-phosphate (Allu6P), and seven D-ribose 5-phosphate isomerase (Rpi) inhibitors. The inhibitors were designed as analogues of the 6-carbon high-energy intermediate postulated for the All6P to Allu6P isomerization reaction (Allpi
Thomas E Edwards et al.
BMC structural biology, 11, 39-39 (2011-10-15)
Ribose-5-phosphate isomerase is an enzyme that catalyzes the interconversion of ribose-5-phosphate and ribulose-5-phosphate. This family of enzymes naturally occurs in two distinct classes, RpiA and RpiB, which play an important role in the pentose phosphate pathway and nucleotide and co-factor
Mirjam M C Wamelink et al.
Journal of molecular medicine (Berlin, Germany), 88(9), 931-939 (2010-05-26)
Ribose 5-phosphate isomerase (RPI) deficiency is an enzymopathy of the pentose phosphate pathway. It manifests with progressive leukoencephalopathy and peripheral neuropathy and belongs, with one sole diagnosed case, to the rarest human disorders. The single patient was found compound heterozygous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service