Skip to Content
Merck
All Photos(2)

Key Documents

P9159

Sigma-Aldrich

Piperidine-4-sulfonic acid

Synonym(s):

P4S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO3S
CAS Number:
Molecular Weight:
165.21
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

form

powder

SMILES string

OS(=O)(=O)C1CCNCC1

InChI

1S/C5H11NO3S/c7-10(8,9)5-1-3-6-4-2-5/h5-6H,1-4H2,(H,7,8,9)

InChI key

UGBJGGRINDTHIH-UHFFFAOYSA-N

Biochem/physiol Actions

GABAA receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Akk et al.
The Journal of physiology, 532(Pt 3), 673-684 (2001-04-21)
Neurosteroids are produced in the brain, and can have rapid actions on membrane channels of neurons. Pregnenolone sulfate (PS) is a sulfated neurosteroid which reduces the responses of the [gamma]-aminobutyric acid A (GABA(A)) receptor. We analysed the actions of PS
B Ebert et al.
Molecular pharmacology, 46(5), 957-963 (1994-11-01)
Using systematic combination of alpha 1, alpha 3, and alpha 5 with beta 1, beta 2, and beta 3, together with gamma 1, gamma 2, and gamma 3, we have investigated the contributions of the various alpha, beta, and gamma
J H Steinbach et al.
The Journal of physiology, 537(Pt 3), 715-733 (2001-12-18)
1. We have studied the kinetic properties of channel gating of recombinant alpha 1 beta 2 gamma 2L GABA(A) receptors transiently expressed in human embryonic kidney 293 cells, using the cell-attached, single-channel patch-clamp technique. The receptors were activated by GABA
J W Miller et al.
Neuroscience, 43(1), 41-49 (1991-01-01)
This study determined the effects of discrete microinjections of GABA agonists in the cholinergic nuclei of the pontomesencephalic tegmentum on spontaneous behavior and seizures induced by intravenous pentylenetetrazol, bicuculline or strychnine, in the rat. Injections of both the GABAA agonist
J W Miller et al.
Neuropharmacology, 29(7), 649-655 (1990-07-01)
This study characterized the role of GABA in the central medial intralaminar nucleus on seizures induced by pentylenetetrazol given systemically. Injections of the direct selective GABAA agonist, piperidine-4-sulfonic acid or the indirect GABAA agonists, flurazepam and pentobarbital, in this region

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service