Skip to Content
Merck
All Photos(1)

Key Documents

524400

Sigma-Aldrich

Phorbol-12-myristate-13-acetate

≥98% (HPLC), solid, PKC activator, Calbiochem®

Synonym(s):

Phorbol-12-myristate-13-acetate, PMA, TPA, PKC Activator I, TPA, PMA, PKC Activator I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Product Name

Phorbol-12-myristate-13-acetate, Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.

Quality Level

description

RTECS - QH4377000

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O([C@]21[C@H]([C@H]3[C@@]([C@H]4[C@](CC(=C3)CO)(C(=O)C(=C4)C)O)([C@@H]([C@H]2OC(=O)CCCCCCCCCCCCC)C)O)C1(C)C)C(=O)C

InChI

1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30-,32-,34-,35-,36-/m1/s1

InChI key

PHEDXBVPIONUQT-RGYGYFBISA-N

General description

Most commonly-used phorbol ester. Extremely potent mouse skin tumor promoter. Activates protein kinase C in vivo and in vitro, even at nM concentrations. Promotes the expression of inducible NOS in cultured hepatocytes. Activates Ca2+-ATPase and potentiates forskolin-induced cAMP formation. Inhibits apoptosis induced by the Fas antigen, but induces apoptosis in HL-60 promyelocytic leukemia cells. Its binding is reversible. Also available as a 10 mM solution in DMSO (Cat. No. 500582 ).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Protein Kinase C
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Anhydrous DMSO stock solutions are stable for up to 6 months at -20°C. With frequent use, the DMSO stock solutions may accumulate water. The presence of water in the stock solution will accelerate the decomposition of the product, resulting in significant loss of activity in 2-3 months.

Other Notes

Powell, D.J., et al. 2003. Mol. Cell Biol.23, 7794.
Tepper, C.G., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8443.
Oishi, K., and Yamaguchi, M. 1994. J. Cell. Biochem. 55, 168.
Macfarlane, D.E., and O’Donnell, P.S. 1993. Leukemia7, 1846.
Ghandi, V.C. and Jones, D.J., 1992. Neuropharmacol.31, 1101.
Hortelano, S., et al. 1992. J. Biol. Chem. 267, 24937.
Kontny, E., et al. 1992. Eur. J. Pharmacol.227, 333.
Zanaboni, P.B., et al. 1992. J. Appl. Physiol.73, 2011.
Saltis, J., et al. 1991. J. Biol. Chem.266, 261.
Beguinot, L., et al. 1985. Proc. Natl. Acad. Sci. USA82, 2774.
Perchellet, J. 1985. Carcinogenesis6, 567.
Nishizuka, Y. 1984. Science255, 1365.
Mastro, A. 1982. Lymphokines6, 263.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gi-Cheon Kim et al.
Frontiers in oncology, 10, 642-642 (2020-06-02)
ETS1 has shown dichotomous roles as an oncogene and a tumor suppressor gene in diverse cancers, but its functionality in breast cancer tumorigenesis still remains unclear. We utilized the Cancer Genome Atlas (TCGA) database to analyze comprehensive functions of ETS1
Ellie Taus et al.
Frontiers in immunology, 13, 835830-835830 (2022-03-12)
CD8+ T cells have key protective roles in many viral infections. While an overall Th1-biased cellular immune response against SARS-CoV-2 has been demonstrated, most reports of anti-SARS-CoV-2 cellular immunity have evaluated bulk T cells using pools of predicted epitopes, without
Nadia Korfali et al.
Methods in molecular biology (Clifton, N.J.), 528, 201-225 (2009-01-21)
The nuclear envelope (NE) is a double membrane system that is both a part of the endoplasmic reticulum and part of the nucleus. As its constituent proteins tend to be highly complexed with nuclear and cytoplasmic components, it is notoriously
Yan-Ruide Li et al.
Cell reports. Medicine, 2(11), 100449-100449 (2021-11-30)
Cell-based immunotherapy has become the new-generation cancer medicine, and "off-the-shelf" cell products that can be manufactured at large scale and distributed readily to treat patients are necessary. Invariant natural killer T (iNKT) cells are ideal cell carriers for developing allogeneic
You Nie et al.
Virology journal, 19(1), 68-68 (2022-04-14)
The onset and progression of cervical intraepithelial neoplasia (CIN) are closely associated with the persistent infection of high-risk HPV (especially type16), which is mainly caused by immune escape. Natural killer (NK) cells play an important role against virally infected cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service