Skip to Content
Merck
All Photos(3)

Key Documents

P5514

Sigma-Aldrich

Pyrilamine maleate salt

Synonym(s):

Mepyramine maleate salt, N-(4-Methoxyphenyl)methyl-N′,N′-dimethyl-N-(2-pyridinyl)-1,2-ethanediamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O · C4H4O4
CAS Number:
Molecular Weight:
401.46
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

form

powder

originator

Watson

SMILES string

OC(=O)\C=C/C(O)=O.COc1ccc(CN(CCN(C)C)c2ccccn2)cc1

InChI

1S/C17H23N3O.C4H4O4/c1-19(2)12-13-20(17-6-4-5-11-18-17)14-15-7-9-16(21-3)10-8-15;5-3(6)1-2-4(7)8/h4-11H,12-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

JXYWFNAQESKDNC-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Pyrilamine maleate salt has been used:
  • to treat Cassiopea sp. as H1 receptor antagonist
  • to intraperitoneally inject mice to prevent the release of endogenous histamine
  • in membrane binding assay to determine the Ki values and in the comparative inhibitory concentration (IC50) studies by whole-cell patch clamp technique

Biochem/physiol Actions

Pyrilamine maleate is a H1 histamine receptor antagonist. Pyrilamine maleate induces sleep in vertebrates. In Cassiopea, it induces concentration dependent dormancy.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Watson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical and biological properties of pyrilamine.
T J Haley
Journal of pharmaceutical sciences, 72(1), 3-12 (1983-01-01)
The [3H] dofetilide binding assay is a predictive screening tool for hERG blockade and proarrhythmia: Comparison of intact cell and membrane preparations and effects of altering [K+] o
Diaz GJ, et al.
Journal of Pharmacological and Toxicological Methods, 50(3), 187-199 (2004)
Evaluating Vascular Hyperpermeability-inducing Agents in the Skin with the Miles Assay
Brash JT, et al.
Journal of Visualized Experiments, 27(136), e57524-e57524 (2018)
S Y Yeh
Synapse (New York, N.Y.), 32(4), 301-311 (1999-05-20)
Various effects of chlorpheniramine (CPA), diphenhydramine (DIPH), tripelennamine (TRIP), and pyrilamine (PYRI) on fenfluramine (FEN)-induced depletion of serotonin in the brain of rats were observed to be dependent on body temperature. Levels of 5-HT and 5-HIAA in the frontal cortex
The jellyfish Cassiopea exhibits a sleep-like state
Nath RD, et al.
Current Biology, 27(19), 2984-2990 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service