Skip to Content
Merck
All Photos(2)

Key Documents

P2767

Sigma-Aldrich

1,2-Di(cis-9-octadecenoyl)-sn-glycerol 3-phosphate sodium salt

≥99% (GC), ≥97% (TLC)

Synonym(s):

L-α-Phosphatidic acid, dioleoyl sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H73O8P
CAS Number:
Molecular Weight:
700.97
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥97% (TLC)
≥99% (GC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].CCCCCCCC\C=C\CCCCCCCC(=O)OCC(COP(O)(O)=O)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/2C39H73O8P.3Na/c2*1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;;;/h2*17-20,37H,3-16,21-36H2,1-2H3,(H2,42,43,44);;;/q;;3*+1/p-3/b2*19-17-,20-18-;;;/t2*37-;;;/m11.../s1

InChI key

VSEVTEXNDGQEBO-LSVXRYHXSA-K

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suttichai Krisanaprakornkit et al.
International immunology, 20(1), 21-29 (2007-11-08)
Human beta-defensin expression correlates with differentiation in oral epithelium, and calcium ion, an important regulator of epithelial differentiation, plays a critical role in regulation of human beta-defensin-2 (hBD-2) mRNA expression. Phospholipase D (PLD) also regulates epithelial differentiation. Therefore, we examined
Luísa Camacho et al.
Journal of cell science, 122(Pt 23), 4383-4392 (2009-11-12)
Rab GTPases of the Arabidopsis Rab-E subclass are related to mammalian Rab8 and are implicated in membrane trafficking from the Golgi to the plasma membrane. Using a yeast two-hybrid assay, Arabidopsis phosphatidylinositol-4-phosphate 5-kinase 2 (PtdIns(4)P 5-kinase 2; also known as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service