Skip to Content
Merck
All Photos(2)

Key Documents

P0639

Sigma-Aldrich

L-α-Phosphatidylinositol sodium salt from Glycine max (soybean)

≥99%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

soybean

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](O)[C@@H](O)[C@@H](OP(OC[C@H](OC([R])=O)COC([R])=O)(O[Na])=O)[C@@H]1O

Biochem/physiol Actions

Phospholipid membrane component, precursor of inositol phosphates.

Other Notes

Contains primarily linoleic and palmitic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongmin Xiong et al.
Journal of cellular physiology, 219(2), 402-414 (2009-01-01)
Previously, we showed that laminin-binding to the dystrophin glycoprotein complex (DGC) of skeletal muscle causes a heterotrimeric G-protein (Galphabetagamma) to bind, changing the activation state of the Gsalpha subunit. Others have shown that laminin-binding to the DGC also leads to
Michael Witting et al.
PloS one, 12(3), e0172311-e0172311 (2017-03-10)
Lipid identification is a major bottleneck in high-throughput lipidomics studies. However, tools for the analysis of lipid tandem MS spectra are rather limited. While the comparison against spectra in reference libraries is one of the preferred methods, these libraries are
S D Tachado et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(8), 4022-4027 (1997-04-15)
The perturbation of various glycosylphosphatidylinositol (GPI)-anchored surface proteins imparts profound regulatory signals to macrophages, lymphocytes and other cell types. The specific contribution of the GPI moieties to these events however is unclear. This study demonstrates that purified GPIs of Plasmodium
Delphine Cardi et al.
The Journal of biological chemistry, 285(34), 26406-26416 (2010-06-10)
The antimalarial drugs artemisinins have been described as inhibiting Ca(2+)-ATPase activity of PfATP6 (Plasmodium falciparum ATP6) after expression in Xenopus oocytes. Mutation of an amino acid residue in mammalian SERCA1 (Glu(255)) to the equivalent one predicted in PfATP6 (Leu) was
Tetsuya Sakurai et al.
Life science alliance, 4(4) (2021-02-13)
DOCK8 is a Cdc42-specific guanine-nucleotide exchange factor that is essential for development and functions of various subsets of leukocytes in innate and acquired immune responses. Although DOCK8 plays a critical role in spatial control of Cdc42 activity during interstitial leukocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service