Skip to Content
Merck
All Photos(2)

Documents

P2611

Sigma-Aldrich

Pectinase from Aspergillus aculeatus

greener alternative

aqueous solution, ≥3,800 units/mL

Synonym(s):

Pectinex® Ultra SPL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

aqueous solution

Quality Level

specific activity

≥3,800 units/mL

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Application

Pectinase is an enzyme from Aspergillus aculeatus that is used in plant protoplast preparation to digest cell wall prior to organelle isolation. It has been used to conduct partial saccharification of sugars. Pectinases are used to study their role in the invasion of plant tissues by phytopathogens, the spoilage of produce and various food processing and plant biotechnology applications. Pectinase, product P2611, has been used to treat leaf discs from fifth wild-type and sitiens leaves.

Biochem/physiol Actions

Pectinase is an active pectolytic enzyme preparation produced by a selected strain of Aspergillus aculeatus that contains mainly pectintranseliminase, polygalacturonase, and pectinesterase and small amounts of hemicellulases and cellulases. Pectinase hydrolyzes pectin, which is a component of the cell wall. They may attack methyl-esterified pectin or de-esterified pectin. It is a source of pectinase activity, also containing cellulase and hemicellulase activities.

Legal Information

A product of Novozyme Corp.
Pectinex is a registered trademark of Novozymes Corp.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Sion et al.
Biotechnology and bioengineering, 118(11), 4453-4464 (2021-08-14)
As a clinical dose requires a minimum of 106 cells per kilogram of patients, it is, therefore, crucial to develop a scalable method of production of Wharton Jelly mesenchymal stem cells (WJ-MSCs) with maintained inner characteristics. Scalable expansion of WJ-MSCs on
Qing Zhang et al.
Frontiers in plant science, 13, 862171-862171 (2022-05-20)
Pectin, cellulose, and hemicellulose constitute the primary cell wall in eudicots and function in multiple developmental processes in plants. Root hairs are outgrowths of specialized epidermal cells that absorb water and nutrients from the soil. Cell wall architecture influences root
Chaowen Xiao et al.
Plant physiology, 170(1), 234-249 (2015-11-04)
Xyloglucan constitutes most of the hemicellulose in eudicot primary cell walls and functions in cell wall structure and mechanics. Although Arabidopsis (Arabidopsis thaliana) xxt1 xxt2 mutants lacking detectable xyloglucan are viable, they display growth defects that are suggestive of alterations
Katrien Curvers et al.
Plant physiology, 154(2), 847-860 (2010-08-17)
A mutant of tomato (Solanum lycopersicum) with reduced abscisic acid (ABA) production (sitiens) exhibits increased resistance to the necrotrophic fungus Botrytis cinerea. This resistance is correlated with a rapid and strong hydrogen peroxide-driven cell wall fortification response in epidermis cells
Sheikh Md Rajiuddin et al.
Frontiers in microbiology, 11, 516445-516445 (2020-10-13)
Consumption of dates has not been considered a common risk of hepatitis A virus (HAV) infection. In January 2018, an outbreak of hepatitis was identified with cases resident in all regions of Denmark. All the detected strains belonged to HAV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service