Skip to Content
Merck
All Photos(2)

Key Documents

P4716

Sigma-Aldrich

Pectinase from Aspergillus niger

greener alternative

BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)

Synonym(s):

Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase solution from Aspergillus niger

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Aspergillus niger

product line

BioReagent

form

aqueous glycerol solution

specific activity

≥5 units/mg protein (Lowry)

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | plant: suitable

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Application

Petctinase from Aspergillus niger has been used:
  • for protoplasts liberation
  • in the protoplast enzyme mixture for enzyme digestion
  • to conduct partial saccharification of sugars
  • to study their role in the invasion of plant tissues by phytopathogens, the spoilage of produce and various food processing and plant biotechnology applications

Used in plant protoplast preparation to digest cell wall prior to organelle isolation.

Biochem/physiol Actions

Pectolytic enzyme preparation produced from a selected strain of Aspergillus niger: contains mainly pectintranseliminase, polygalacturonase, and pectinesterase and small amounts of hemicellulases and cellulases. Pectinases hydrolyses pectin, which is a component of the cell wall. They may attack methyl-esterified pectin or de-esterified pectin. It is a source of pectinase activity, also containing cellulase and hemicellulase activities.Pectinase catalyzes the random hydrolysis of a-(1-4)-Dgalactosiduronic linkages in pectin and other galacturonans.

Unit Definition

One unit will liberate 1.0 μmole of galacturonic acid from polygalacturonic acid per min at pH 4.0 at 25 °C.

Physical form

Solution in 40% glycerol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karyotypes of some species of Castanopsis, Lithocarpus and Quercus (Fagaceae) from Khun Mae Kuang Forest in Chiang Mai province, northern Thailand
Chokchaichamnankit P, et al.
Thai Forest Bulletin , 38-44 (2007)
Tony Heitkam et al.
The Plant journal : for cell and molecular biology, 103(1), 32-52 (2020-01-26)
If two related plant species hybridize, their genomes may be combined and duplicated within a single nucleus, thereby forming an allotetraploid. How the emerging plant balances two co-evolved genomes is still a matter of ongoing research. Here, we focus on
Jason Q D Goodger et al.
PloS one, 11(3), e0151432-e0151432 (2016-03-16)
The sub-dermal secretory cavities (glands) embedded within the leaves of Eucalyptus (Myrtaceae) were once thought to be the exclusive repositories of monoterpene and sesquiterpene oils. Recent research has debunked this theory and shown that abundant non-volatile compounds also occur within
Tolerance mechanisms in mercury-exposed Chromolaena odorata (lf) RM King et H. Robinson, a potential phytoremediator
Alcantara HJP, et al.
Journal of Degraded and Mining Lands Management, 1(1), 09-20 (2013)
Trude Schwarzacher
Methods in molecular biology (Clifton, N.J.), 1370, 87-103 (2015-12-15)
Good preparations are essential for informative analysis of both somatic and meiotic chromosomes, cytogenetics, and cell divisions. Fluorescent chromosome staining allows even small chromosomes to be visualized and counted, showing their morphology. Aneuploidies and polyploidies can be established for species

Protocols

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service