Skip to Content
Merck
All Photos(1)

Key Documents

P134

Sigma-Aldrich

(−)-Pentazocine

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27NO
CAS Number:
Molecular Weight:
285.42
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

drug control

USDEA Schedule IV; Home Office Schedule 3; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

storage condition

protect from light

color

white

solubility

DMSO: >10 mg/mL

originator

Sanofi Aventis

storage temp.

room temp

SMILES string

C[C@@H]1[C@@H]2Cc3ccc(O)cc3[C@@]1(C)CCN2C\C=C(\C)C

InChI

1S/C19H27NO/c1-13(2)7-9-20-10-8-19(4)14(3)18(20)11-15-5-6-16(21)12-17(15)19/h5-7,12,14,18,21H,8-11H2,1-4H3/t14-,18+,19+/m1/s1

InChI key

VOKSWYLNZZRQPF-CCKFTAQKSA-N

Biochem/physiol Actions

Pentazocine is an analgesic with both agonist and antagonist activities. It acts as a κ- and σ-opioid receptor agonist and partial mu-receptor antagonist. Pentazocine pre-treatment reduces the frequency of fentanyl-induced cough and improves the anesthetic effect.[1]

Features and Benefits

This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Grosse Maestrup et al.
Bioorganic & medicinal chemistry, 19(1), 393-405 (2010-12-04)
In order to develop a fluorinated radiotracer for imaging of σ(1) receptors in the central nervous system a series of (2-fluoroethyl) substituted spirocyclic piperidines 3 has been prepared. In the key step of the synthesis 2-bromocinnamaldehyde acetal 5 was added
Carmen Abate et al.
Journal of medicinal chemistry, 54(4), 1022-1032 (2011-01-15)
1-Cyclohexyl-4-[3-(5-methoxy-1,2,3,4-tetrahydronaphthalen-1-yl)propyl]piperazine 1 (PB28) represents an excellent lead candidate for therapeutic and/or diagnostic applications in oncology. However, because its utility is limited by its relatively high degree of lipophilicity, novel analogues of 1 with reduced lipophilic character were designed by substituting
Christian Geiger et al.
Journal of medicinal chemistry, 50(24), 6144-6153 (2007-10-31)
All possible stereoisomeric alcohols (6-benzyl-8-(4-methoxybenzyl)-6,8-diazabicyclo[3.2.2]nonan-2-ol) and methyl ethers (6-benzyl-2-methoxy-8-(4-methoxybenzyl)-6,8-diazabicyclo[3.2.2]nonane) are prepared from (R)- and (S)-glutamate. A Dieckmann analogous cyclization, which makes use of trapping the primary cyclization product with Me3SiCl, generates the bicyclic framework. Stereoselective LiBH4 reduction and Mitsunobu inversion
Qing Ai et al.
Pharmacological reports : PR, 62(4), 747-750 (2010-10-05)
This study evaluated the effect of pentazocine pretreatment on fentanyl-induced cough. With ethics committee approval, 277 ASA I-II patients, aged between 19 and 63 years, undergoing various elective surgeries during general anesthesia, were enrolled in this prospective, randomized, double-blind, placebo-controlled
Daniele Zampieri et al.
Journal of medicinal chemistry, 52(17), 5380-5393 (2009-08-14)
Novel benzo[d]oxazol-2(3H)-one derivatives were designed and synthesized, and their affinities against sigma receptors were evaluated. On the basis of 31 compounds, a three-dimensional pharmacophore model for the sigma(1) receptor binding site was developed using the Catalyst 4.9 software package. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service