Skip to Content
Merck
All Photos(3)

Documents

P1263

Sigma-Aldrich

1,2-Didodecanoyl-sn-glycero-3-phosphocholine

≥99%, synthetic

Synonym(s):

PC, (7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxododecyl)oxy]-3,5,9-trioxa-4-phosphaheneicosan-1-aminium 4-oxide, inner salt, 1,2-Dilauroyl-sn-glycero-3-phosphocholine, 1,2-Dilauroyl-sn-glycero-3-phosphorylcholine, 3-sn-Phosphatidylcholine, 1,2-dilauroyl, L-β,γ-Dilauroyl-α-lecithin, DLPC, PC(12:0/12:0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C32H64NO8P
CAS Number:
Molecular Weight:
621.83
Beilstein:
5676925
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O)=O

InChI

1S/C32H64NO8P/c1-6-8-10-12-14-16-18-20-22-24-31(34)38-28-30(29-40-42(36,37)39-27-26-33(3,4)5)41-32(35)25-23-21-19-17-15-13-11-9-7-2/h30H,6-29H2,1-5H3/t30-/m1/s1

InChI key

IJFVSSZAOYLHEE-SSEXGKCCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Papandreou et al.
Nutrients, 11(5) (2019-05-11)
Few studies have examined the association of a wide range of metabolites with total and subtypes of coffee consumption. The aim of this study was to investigate associations of plasma metabolites with total, caffeinated, and decaffeinated coffee consumption. We also
Katharine Hammond et al.
ACS infectious diseases, 5(8), 1471-1479 (2019-05-24)
A design template for membrane active antibiotics against microbial and tumor cells is described. The template is an amino acid sequence that combines the properties of helminth defense molecules, which are not cytolytic, with the properties of host-defense peptides, which
Plasma lipidomic profiles and cardiovascular events in a randomized intervention trial with the Mediterranean diet.
Estefanía Toledo et al.
The American journal of clinical nutrition, 106(4), 973-983 (2017-08-18)
Cecilia Pinto et al.
Journal of structural biology, 206(1), 1-11 (2017-12-05)
Significant progress has been made in obtaining structural insight into the assembly of the β-barrel assembly machinery complex (BAM). These crystallography and electron microscopy studies used detergent as a membrane mimetic and revealed structural variations in the central domain, BamA
Nina Østergaard Knudsen et al.
International journal of pharmaceutics, 416(2), 478-485 (2011-03-23)
Many dermal diseases like psoriasis are characterized by major changes in skin barrier function, which challenge the reproducible delivery of drugs into specific layers of diseased skin. The purpose of this study was to elucidate how liposomal bilayer fluidity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service