Skip to Content
Merck
All Photos(1)

Key Documents

P0060

Sigma-Aldrich

PLK1, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

STPK13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

14-20 nmol/min·mg

mol wt

~70 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PLK1(5347)

General description

PLK1 (polo like kinase 1) gene is mapped to human chromosome 16p12.2. It codes for the protein serine/threonine kinase.

Biochem/physiol Actions

PLK1 (polo like kinase 1) is predominantly involved in mitosis. It mediates the phosphorylation of cyclin B1/CDK1 (cyclin dependent kinase 1) complex, and activation of the anaphase promoting complex. PLK1 has anti-apoptotic function. Upregulation of gene is observed in different types of cancer, such as colorectal cancer and is regarded as an oncogene. PLK1 is involved in checkpoint recovery and adaptation following DNA damage.

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polo-like kinase 1 expression is suppressed by CCAAT/enhancer-binding protein a to mediate colon carcinoma cell differentiation and apoptosis.
Dasgupta N
Biochimica et Biophysica Acta (2017)
Wei Dai et al.
Progress in cell cycle research, 5, 327-334 (2003-11-05)
Studies from eukaryotic model systems, ranging from yeast to human, indicate that Polo and Polo-like kinases (Plks) are essential for the activity of the microtubule organization center. Polo/Plks localize to centrosomes or spindle pole bodies and undergo dramatic subcellular relocation
Polo-like Kinase 1 as a potential therapeutic target in Diffuse Intrinsic Pontine Glioma.
Amani V
BMC Cancer (2016)
PLK1 inhibition enhances temozolomide efficacy in IDH1 mutant gliomas.
Koncar RF
Oncotarget (2017)
Expression of the microtubule-associated protein MAP9/ASAP and its partners AURKA and PLK1 in colorectal and breast cancers.
Rouquier S
Disease Markers (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service