Skip to Content
Merck
All Photos(1)

Key Documents

N9784

Sigma-Aldrich

Neutralization Solution for Blood

sufficient for 100 reactions, sufficient for 1000 reactions, for molecular biology

Synonym(s):

DNA neutralization solution, Neutralization buffer DNA extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

liquid

usage

sufficient for 100 reactions
sufficient for 1000 reactions

storage temp.

room temp

General description

Neutralization Solution for Blood is used along with lysis solution for blood (L3289) for the rapid extraction and neutralization of human genomic DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

This solution is a component of the Extract-N-Amp and the REDExtract-N-Amp Blood PCR Kits

Application

Neutralization Solution for blood has been used to neutralize extracted DNA prior to polymerase chain reaction (PCR) amplification.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Veith et al.
International journal of chronic obstructive pulmonary disease, 14, 2535-2542 (2019-12-11)
Alpha-1-antitrypsin deficiency (AATD) is a rare hereditary condition resulting from the mutations in the SERPINA1 (serine protease inhibitor) gene and is characterized by low circulating levels of the alpha-1 antitrypsin (AAT) protein. The traditional algorithm for laboratory testing of AATD
Leandro S Thiago et al.
Haematologica, 99(1), 155-162 (2013-07-23)
The B-cell compartment in which multiple myeloma stem cells reside remains unclear. We investigated the potential presence of mature, surface-membrane immunoglobulin-positive B lymphocytes clonally related to the tumor bone marrow plasma cells among different subsets of peripheral blood B cells
G Henderson et al.
Archives of disease in childhood. Fetal and neonatal edition, 94(2), F124-F128 (2007-09-05)
The inflammatory cytokine cascade is implicated in the pathogenesis of necrotising enterocolitis (NEC). Genetic association studies of cytokine polymorphisms may help to detect molecular mechanisms that are causally related to the disease process. To examine associations between the common genetic
Huiling Wu et al.
The Journal of clinical investigation, 117(10), 2847-2859 (2007-09-15)
Ischemia/reperfusion injury (IRI) may activate innate immunity through the engagement of TLRs by endogenous ligands. TLR4 expressed within the kidney is a potential mediator of innate activation and inflammation. Using a mouse model of kidney IRI, we demonstrated a significant
Aaron Zhong et al.
STAR protocols, 1(2) (2020-10-20)
Reporter cell lines based on human pluripotent stem cells (hPSCs) are highly desirable for studying differentiation, lineage tracing, and target cell selection. However, several technical bottlenecks, such as DNA transduction, low homology recombination rate (HDR), and single-cell cloning, have made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service