Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1303Z

Sigma-Aldrich

MDCKII canine MDR1 KO, human MDR1 Expressing Cells

Synonym(s):

MDCKII cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.81

biological source

canine kidney (cocker spaniel)

usage

sufficient for 1 96-well plate(s) (or 24-well plate)

packaging

vial of 2 million cells

growth mode

adherent

technique(s)

cell culture | mammalian: suitable
drug transporter assay: suitable

application(s)

ADME/TOX

shipped in

dry ice

storage temp.

−196°C

General description

MDCKII - Madin-Darby canine kidney - is a subclone derived from the heterogenous parent line MDCK (ECACC Catalog No. 85011435). MDCKII cells, which predominate in later passages from MDCK, are reported to display electrical resistance of 100 ohm/cm2. This cell line is thought to be derived from the distal tubule or collecting duct of the nephron. The cell line can be used as an experimental model to study the generation and maintenance of cell surface polarity in epithelial cells.

Application

ATP-binding cassette (ABC) transporters are a family of transmembrane proteins that utilize ATP hydrolysis for translocation of substrates across membranes. ABC transporters are known to play a critical role in the development of multidrug resistance. Evaluation of membrane transporter pharmacology in drug disposition and drug-drug interactios (DDI) is critical to the pharmaceutical safety evaluations of new drug entities. Selection of the targeted gene(s) was based on the considerable body of evidence supporting its crucial role in the development of multidrug resistance.

These cells can be used in transwell permeability assays to measure the movement of substrates through the monolayer. Efflux ratios for test compounds can be determined using standard compounds as controls.

Features and Benefits

CompoZr® zinc finger nuclease (ZFN) technology is a fast and reliable way to manipulate the genome in a targeted fashion. ZFNs are naturally occurring proteins that can be engineered to bind DNA at a sequence-specific location and create a double strand break. The cell′s natural machinery repairs the break in one of two ways: non-homologous end joining or homologous recombination. The non-homologous end joining pathway typically produces small modifications (indels) at the targeted locus that may result in a functional knockout. Single cell clones are then isolated, tested for the desired modification and expanded to establish a stable cell line.

The canine MDR1 (cP-gp) efflux transporter gene has been effectively disrupted in both alleles. There is no expression of the cP-gp. Validation studies have shown no efflux of standard cP-gp substrates.

The human MDR1 efflux transporter has been expressed in these cells via viral transduction. Validation studies have shown efflux ratios ≥2 for common MDR1 substrates digoxin, erythromycin, and quinidine.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification

Legal Information

These products are covered by the ADME/Tox Cell Lines License as described in Exhibit 1, in the technical bulletin.

MDCKII sublone was originally isolated by Daniel Louvard, Institut Curie, Paris France.
References: Hansson, G.G., Simons, K and Van Meer G (1986) EMBO 5: 483-489, Louvard D (1980) PNAS 77; 4132-4136
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deep Agnani et al.
PloS one, 6(10), e25086-e25086 (2011-10-27)
P-glycoprotein, a human multidrug resistance transporter, has been extensively studied due to its importance to human health and disease. In order to understand transport kinetics via P-gp, confluent cell monolayers overexpressing P-gp are widely used. The purpose of this study
S Simon et al.
Biochimica et biophysica acta, 1821(9), 1211-1223 (2012-06-14)
Phospholipids are widely used excipients for pharmaceutical formulations, such as for preparing biphasic systems or to solubilize or encapsulate poorly soluble drugs. The present study investigates a new property of this class of substance: its ability to inhibit the efflux

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service