Skip to Content
Merck
All Photos(3)

Key Documents

MBD0015

Sigma-Aldrich

DAPI ready made solution

DAPI ready made solution

For Nuclear counterstain in immunoflourecence microscopy, High Content Screening (HCS), Chromosome staining and flow cytometry (FACS)., 1 mg/mL

Synonym(s):

4′,6-Diamidino-2-phenylindole dihydrochloride, DAPI Dye, DAPI Stain, DNA Content Counterstain, 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, DAPI dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N5 · 2HCl
CAS Number:
Molecular Weight:
350.25
Beilstein:
4894417
MDL number:
UNSPSC Code:
41105500
NACRES:
NA.55
Pricing and availability is not currently available.

Quality Level

form

liquid

storage condition

protect from light

concentration

1 mg/mL

technique(s)

transfection: suitable

solubility

water: soluble

fluorescence

λex 340 nm; λem 488 nm (nur DAPI)
λex 364 nm; λem 454 nm (DAPI-DNA-Komplex)

suitability

suitable for nucleic acid staining

shipped in

dry ice

storage temp.

−20°C

SMILES string

Cl.Cl.NC(=N)c1ccc(cc1)-c2cc3ccc(cc3[nH]2)C(N)=N

InChI

1S/C16H15N5.2ClH/c17-15(18)10-3-1-9(2-4-10)13-7-11-5-6-12(16(19)20)8-14(11)21-13;;/h1-8,21H,(H3,17,18)(H3,19,20);2*1H

InChI key

FPNZBYLXNYPRLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DAPI, 4′,6-diamidino-2-phenylindole, is a blue fluorescent nucleic acid stain that fluoresces brightly upon selectively binding to the minor groove of double stranded DNA. Its selectivity for DNA and high cell permeability allows efficient staining of nuclei with little background from the cytoplasm. The excitation and emission wavelengths of DAPI-DNA complex are 360 nm and 460 nm, respectively.

Application

DAPI Readymade Solution is provided at 1mg/mL concentration. It is recommended to dilute DAPI readymade solution 1:1000 with a desired buffer for a 1μg/mL working concentration (typical working concentration used in most applications is 0.1μg/mL-10μg/mL). Store DAPI solution (1mg/mL) -20°C protected from light. It is recommended to aliquot DAPI readymade solution and to avoid freeze thaw cycles.

DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma.

For Nuclear counterstain in immunoflourecence microscopy, High Content Screening (HCS), Chromosome staining and flow cytometry (FACS).
DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels.
DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma.

Biochem/physiol Actions

Cell permeable fluorescent minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA (preferentially to AT rich DNA), forming a stable complex which fluoresces approximately 20 times greater than DAPI alone.

Features and Benefits

  • Ready-to-use liquid, cell permeable dye.
  • Can be used with live cells and fixed cells.
  • Effective for fixed-cell staining and quantitation of DNA content.
  • Optimal Counterstain when using fluorescent antibodies during fluorescence microscopy or high-content screening (HCS) applications.

Physical form

DAPI readymade solution is provided at 1mg/mL concentration in water

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

June Zhang et al.
Chemosphere, 256, 127038-127038 (2020-05-30)
Baicalein is a flavonoid that is widely found in plants. Studies have shown that baicalein has anti-inflammatory, anti-cancer, and liver-protective effects. However, the effects of baicalein on TAA-induced toxicity and the underlying molecular mechanisms in zebrafish larvae are still unknown.
Mark Jones et al.
Cell transplantation, 29, 963689720923578-963689720923578 (2020-07-15)
Absence or reduced frequency of human regulatory T cells (Tregs) can limit the control of inflammatory responses, autoimmunity, and the success of transplant engraftment. Clinical studies indicate that use of Tregs as immunotherapeutics would require billions of cells per dose.
Maksim A Nesterenko et al.
Parasites & vectors, 13(1), 559-559 (2020-11-11)
Parasitic flatworms (Trematoda: Digenea) represent one of the most remarkable examples of drastic morphological diversity among the stages within a life cycle. Which genes are responsible for extreme differences in anatomy, physiology, behavior, and ecology among the stages? Here we report
Wen Zeng et al.
Frontiers in cellular neuroscience, 15, 638351-638351 (2021-03-09)
Brain microvascular endothelial cells (BMECs) are a major component of the blood-brain barrier that maintains brain homeostasis. Preserving and restoring the normal biological functions of BMECs can reverse or reduce brain injury. Endothelial progenitor cells (EPCs) may promote brain vascular
Ting-Ting Yu et al.
Cell & bioscience, 11(1), 47-47 (2021-03-04)
Skeletal development and maintenance are complex processes known to be coordinated by multiple genetic and epigenetic signaling pathways. However, the role of long non-coding RNAs (lncRNAs), a class of crucial epigenetic regulatory molecules, has been under explored in skeletal biology.

Reviews

Active Filters

  1. Egypt
    • Review 1
    • Votes 0
    1 out of 5 stars.

    The product is not working on frozen sections

    I bought this product to stain frozen sections however the product did not stain frozen sections. The R and D team did not have specific recommendation.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service