Skip to Content
Merck
All Photos(1)

Key Documents

F4680

Sigma-Aldrich

Fluoromount Aqueous Mounting Medium

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

Product Name

Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues

grade

for fluorescence

Quality Level

form

liquid

technique(s)

microbe id | staining: suitable

color

clear

suitability

suitable for immunohistochemistry

application(s)

diagnostic assay manufacturing
hematology
histology

General description

Fluoromount is an aqueous based mounting medium,similar to Gel Mountä, for use with coverslips to preserve tissues stained with fluorescent dyes. Thisproduct is compatible with FITC, phycoerythrin, phycocyanin, and allophycocyanin labeling. It can alsobe used with other fluorescent markers such as TexasRedÒand rhodamine. The medium after use can easily be removed by soaking the slides in deionized water.

Application

Fluoromount Aqueous Mounting Medium has been used for mounting:
  • coverslips on slides for immunofluorescence microscopic examination of PC12 and Hela cells
  • hippocampus neurons cells for confocal microscope studies
  • brain sections for immunofluorescence staining

Legal Information

Fluoromount is a trademark of Diagnostic BioSystems, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biophysics of microchannel-enabled neuron-electrode interfaces
Wang L, et al.
Journal of Neural Engineering, 9(2), 026010-026010 (2012)
David Fast et al.
mBio, 9(4) (2018-08-02)
Adult Drosophila melanogaster raised in the absence of symbiotic bacteria have fewer intestinal stem cell divisions and a longer life span than their conventionally reared counterparts. However, we do not know if increased stem cell divisions are essential for symbiont-dependent
Kyle J Martin et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(51), 14769-14774 (2016-12-09)
Teeth and denticles belong to a specialized class of mineralizing epithelial appendages called odontodes. Although homology of oral teeth in jawed vertebrates is well supported, the evolutionary origin of teeth and their relationship with other odontode types is less clear.
The role of chaperone-mediated autophagy in huntingtin degradation
Qi L, et al.
Testing, 7(10), e46834-e46834 (2012)
Mohammed Oufattole et al.
Endocrinology, 147(5), 2138-2146 (2006-02-04)
IGF-binding protein (IGFBP)-3 has intrinsic antiproliferative and proapoptotic functions that are independent of IGF binding and may involve nuclear localization. We determined that exogenous IGFBP-3 rapidly translocates to myoblast nuclei and that a 22-residue peptide containing the metal binding domain

Articles

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service