Skip to Content
Merck
All Photos(1)

Key Documents

M7891

Supelco

N-Methyl-N-(trimethylsilyl)trifluoroacetamide

BioReagent, for silylations, LiChropur

Synonym(s):

N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)3
CAS Number:
Molecular Weight:
199.25
Beilstein:
1941550
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.25

vapor density

>1 (vs air)

vapor pressure

8.8 mmHg ( 27 °C)

product line

BioReagent

form

liquid

quality

for silylations, LiChropur

reaction suitability

reaction type: Silylations
reagent type: derivatization reagent

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.38 (lit.)

bp

130-132 °C (lit.)

solubility

chloroform: soluble 0.1 mg/mL

density

1.07 g/mL at 25 °C (lit.)
1.075 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-N-(trimethylsilyl)trifluoroacetamide(MSTFA) is a BioReagent commonly used for silylation reactions in analyticalchemistry.

Application

N-Methyl-N-(trimethylsilyl)trifluoroacetamide can be used:
  • In the derivatization step of the metabolomics analysis using gas chromatography/mass spectrometry (GC/MS).
  • Derivatization reagent in gas chromatography with flame ionization detection (GC-FID) analysis.

Packaging

Sealed ampules contain 1 ml. Gram sizes are in screw-cap bottles.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Response of Downy Oak (Quercus pubescens Willd.) to climate change: Transcriptome assembly, differential gene analysis and targeted metabolomics
Mevy JP, et al.
Plants (Basel, Switzerland), 9(9), 1149-1149 (2020)
A gas chromatography-mass spectrometry-based whole-cell screening assay for target identification in distal cholesterol biosynthesis
Muller C, et al.
Nature Protocols, 14(8), 2546-2570 (2019)
Stina K Lien et al.
Journal of chromatography. A, 1247, 118-124 (2012-06-15)
GC-MS analysis of silylated metabolites is a sensitive method that covers important metabolite groups such as sugars, amino acids and non-amino organic acids, and it has become one of the most important analytical methods for exploring the metabolome. Absolute quantitative
Hee-Kyung Jeon et al.
Journal of chromatography. A, 1131(1-2), 192-202 (2006-08-08)
A novel method has been developed to simultaneously determine and quantify seven organic UV filters employing liquid (solid)-liquid extraction, derivatization with N-methyl-N-(trimethylsilyl) trifluoroacetamide (MSTFA) and gas chromatography with mass spectrometric detection in various environmental matrices. The UV filters determined were:
Iria González-Mariño et al.
Journal of chromatography. A, 1217(11), 1748-1760 (2010-02-13)
An alternative method for the sensitive determination of several drugs of abuse and some of their metabolites in surface and sewage water samples is proposed. Analytes are concentrated using a solid-phase extraction (SPE) sorbent, converted into the corresponding trimethylsilyl derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service