Skip to Content
Merck
All Photos(2)

Key Documents

8.05968

Sigma-Aldrich

N,N′-Methylenediacrylamide

for synthesis

Synonym(s):

N,N′-Methylenediacrylamide, BIS, Bisacrylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352111
EC Index Number:
203-750-9
NACRES:
NA.22
Assay:
≥98%

Quality Level

Assay

≥98%

form

solid

potency

390 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: C-C Bond Formation

pH

≥5.0 (20 °C, 25 g/L in H2O)

mp

185 °C (decomposition)

density

1.235 g/cm3 at 30 °C

bulk density

200 kg/m3

storage temp.

2-30°C

SMILES string

N(CNC(=O)C=C)C(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Application

  • One step photopolymerization of N, N-methylene diacrylamide: This study explores the use of N,N′-Methylenediacrylamide in the photopolymerization process to enhance the antibacterial performance of carboxymethyl starch-silver nanoparticles on cotton fabrics. (El-Sheikh and Ibrahim, 2014).
  • Ultra-stretchable and force-sensitive hydrogels: Research involving the reinforcement of hydrogels with chitosan microspheres embedded in polymer networks using N,N′-Methylenediacrylamide, highlighting its application in creating durable and flexible materials. (Duan et al., 2016).
  • Gamma radiation induced preparation of composite material: A study detailing the use of N,N′-Methylenediacrylamide in the synthesis of organic-inorganic composite materials for the sorption of cesium and zinc. (Allan et al., 2014).
  • Development of thermo-sensitive and magnetic molecularly imprinted polymer: This research focuses on the extraction of omeprazole from biological and pharmaceutical samples, utilizing a polymer network involving N,N′-Methylenediacrylamide. (Beheshti et al., 2023).

Analysis Note

Assay (ex N): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service