Skip to Content
Merck
All Photos(1)

Key Documents

LO4100

Sigma-Aldrich

LOPAC®1280 - Small Scale

Synonym(s):

Library of Pharmacologically Active Compounds - small scale

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

form

liquid

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Compounds are arranged in 96-well format in 16 racks of 80, one compound per well (25 μl at 10 mM in DMSO in archival storage racks).
LOPAC® 1280 is an extensively used library of pharmacologically active compounds. This library includes 1,280 marketed drugs, failed development candidates, and small molecules with well-characterized activities. LOPAC 1280 spans a broad molecular weight range, exhibits multiple mechanisms of action and interacts with a variety of biological receptors.

Application

LOPAC® 1280 - Small Scale has been used:
  • in sprouting assay to screen potential lymphangiogenic inhibitors and the signaling pathways involved in lymphangiogenesis
  • for identification of neuroactive compounds by measuring spontaneous activity within single zebrafish embryos by high-content screening assay
  • for high-throughput Screen for ectonucleoside triphosphate diphosphohydrolase-1 (ENTPD1; also known as CD39) and ecto-5′-nucleotidase (CD73) inhibitors

Features and Benefits

Includes the latest, drug-like molecules in the fields of Cell Signaling & Neuroscience:

  • Apoptosis
  • G Proteins & Cyclic Nucleotides
  • Gene Regulation & Expression
  • Ion Channels
  • Lipid Signaling
  • Multi-Drug Resistance
  • Neurotransmission
  • Phosphorylation
SDFile provided to construct a database containing:
  • Structure
  • Primary Name
  • Secondary Name
  • Pharmacological Activity
  • Product Number
  • Rack Position

Other Notes

For sale inside of the U.S. only. Customers outside the US should refer to Product Code LO4200.

Legal Information

LOPAC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Lact. - Muta. 1B - Repr. 1A - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland E Kälin et al.
Blood, 114(5), 1110-1122 (2009-05-30)
Angiogenesis and lymphangiogenesis are essential for organogenesis but also play important roles in tissue regeneration, chronic inflammation, and tumor progression. Here we applied in vivo forward chemical genetics to identify novel compounds and biologic mechanisms involved in (lymph)angiogenesis in Xenopus
Sara M Vliet et al.
Toxicology and applied pharmacology, 329, 241-248 (2017-06-18)
Spontaneous activity represents an early, primitive form of motor activity within zebrafish embryos, providing a potential readout for identification of neuroactive compounds. However, despite use as an endpoint in chemical screens around the world, the predictive power and limitations of
Miao Xu et al.
Nature medicine, 22(10), 1101-1107 (2016-08-30)
In response to the current global health emergency posed by the Zika virus (ZIKV) outbreak and its link to microcephaly and other neurological conditions, we performed a drug repurposing screen of ∼6,000 compounds that included approved drugs, clinical trial drug
Martin Michael Peter Schulz et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(40), E2665-E2674 (2012-09-06)
Lymphangiogenesis plays an important role in promoting cancer metastasis to sentinel lymph nodes and beyond and also promotes organ transplant rejection. We used human lymphatic endothelial cells to establish a reliable three-dimensional lymphangiogenic sprouting assay with automated image acquisition and
Meera Kumar et al.
SLAS discovery : advancing life sciences R & D, 25(3), 320-326 (2019-12-24)
Production of adenosine in the extracellular tumor microenvironment elicits strong immunosuppression and is associated with tumor progression. Thus, targeting adenosine-generating ectonucleotidases is a potential strategy to stimulate and prolong antitumor immunity. Because the reaction products of ectonucleotidases differ by a

Articles

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Explore compound library screening options with our Pharmacologically Active Compounds portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service