L6512
Lectin from Helix pomatia
biotin conjugate, lyophilized powder
Synonym(s):
Helix pomatia agglutinin, HPA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
conjugate
biotin conjugate
form
lyophilized powder
extent of labeling
2-4 mol biotin per mol protein
impurities
salt, free
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Lectins are carbohydrate-binding proteins, omnipresent, found in fungi, plants and animals. These oligomeric proteins are characterized by the presence of a carbohydrate recognition domain.
Application
Lectin from Helix pomatia has been used to study its binding activities in Gyrodactylus derjavini parasitizing fins of Oncorhynchus mykiss.
Biochem/physiol Actions
HPA has anti-A human blood group specificity and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.
Lectin is useful in glycoconjugate characterizing, imaging and targeting. Lectin participates in host recognition and tissue adhesion. Lectins shows resistance to proteolytic degradation in vivo.
Analysis Note
Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hour incubation at 25 °C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Histochemical characteristics of Gyrodactylus derjavini parasitizing the fins of rainbow trout (Oncorhynchus mykiss)
Buchmann K, et al.
Folia Parasitol (Praha), 45(4), 312-318 (1998)
Overview of fish immunity
Mucosal Health in Aquaculture, 3-54 (2015)
Cédric Jäger et al.
BMC nephrology, 23(1), 178-178 (2022-05-11)
Recurrence of IgA nephropathy (IgAN) after kidney transplantation occurs in about 30% of patients. The relevance of recurrence for the long-term graft survival is expected to increase, since graft survival continues to improve. In a nested study within the Swiss
PEAS AND LENTILS
Encyclopedia of food sciences and nutrition, 4433-4440 (2003)
Yoko Nabeshima et al.
Scientific reports, 10(1), 19122-19122 (2020-11-07)
Human group-specific component protein (Gc protein) is a multifunctional serum protein which has three common allelic variants, Gc1F, Gc1S and Gc2 in humans. Gc1 contains an O-linked trisaccharide [sialic acid-galactose-N-acetylgalactosamine (GalNAc)] on the threonine420 (Thr420) residue and can be converted
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service