Skip to Content
Merck
All Photos(1)

Key Documents

L1516

Sigma-Aldrich

Lectin from Wisteria floribunda

buffered aqueous solution, biotin conjugate

Synonym(s):

Wisteria floribunda agglutinin, WFA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

conjugate

biotin conjugate

Quality Level

form

buffered aqueous solution

packaging

pkg of 2 mg active conjugate

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

WFA is not blood group specific, but has an affinity for N-acetyl-D-galactosamine.

Packaging

Package size based on active conjugate content

Preparation Note

Protein is biotinylated utilizing aminocaproyl spacer.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Mennesson et al.
eNeuro, 7(4) (2020-08-14)
NETO2 is an auxiliary subunit for kainate-type glutamate receptors that mediate normal cued fear expression and extinction. Since the amygdala is critical for these functions, we asked whether Neto2 -/- mice have compromised amygdala function. We measured the abundance of
Margaret Po-Shan Luke et al.
Molecular and cellular neurosciences, 107, 103527-103527 (2020-07-08)
Vision loss has long since been considered irreversible after a critical period; however, there is potential to restore limited vision, even in adulthood. This phenomenon is particularly pronounced following complete loss of vision in the dominant eye. Adult neural cell
Jenni Kyyriäinen et al.
Epilepsy research, 151, 67-74 (2019-03-06)
Extracellular proteolysis initiated by the binding of urokinase-type plasminogen activator (uPA) to its receptor (uPAR) regulates the development of inhibitory neuronal circuits in the cerebral cortex and tissue remodeling after epileptogenic brain injury. To study the function of different components
Natalia Uriarte et al.
Scientific reports, 10(1), 5479-5479 (2020-03-28)
Perineuronal nets (PNNs) are aggregations of extracellular matrix associated with specific neuronal populations in the central nervous system, suggested to play key roles in neural development, synaptogenesis and experience-dependent synaptic plasticity. Pregnancy and lactation are characterized by a dramatic increase
Kelly E Carstens et al.
The European journal of neuroscience, 53(12), 4005-4015 (2020-11-22)
Current methods of experimentally degrading the specialized extracellular matrix (ECM), perineuronal nets (PNNs) have several limitations. Genetic knockout of ECM components typically has only partial effects on PNNs, and knockout of the major ECM component aggrecan is lethal in mice.

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service