Skip to Content
Merck
All Photos(1)

Key Documents

L0543

Sigma-Aldrich

LB Agar Kanamycin-50, Plates

pre-poured LB agar plates with 50 ug/ml kanamycin

Synonym(s):

kanamycin agar plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

grade

for molecular biology

sterility

sterile; autoclaved

form

plates (10 cm)
plates (10cm, 20-25ml each)

capacity

20-25 mL

storage temp.

2-8°C

suitability

selective for Escherichia coli
selective for coliforms

General description

Miller′s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.

Application

Suitable for selective cultivation of E. coli strains that contain plasmids conferring kanamycin resistance. The bacteria can be used for DNA plasmid production or production of recombinant proteins. LB Agar Kanamycin-50, Plates has been used in the screening of Agrobacterium tumefaciens and E.coli JM83 strain with pGD103 plasmid.

Features and Benefits

Miller′s LB Agar plates provide:
  • Convenient, ready-to-use 10cm format
  • Standard formulation plus antibiotic

Components

15g/L Agar
10g/L Tryptone
10g/L NaCl
5g/L Yeast Extract
50μg/ml kanamycin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agrobacterium-mediated transformation and insertional mutagenesis in Colletotrichum acutatum for investigating varied pathogenicity lifestyles
Talhinhas P, et al.
Molecular Biotechnology, 39(1), 57-67 (2008)
Cloning of genes encoding a 15,000-dalton peptidoglycan-associated outer membrane lipoprotein and an antigenically related 15,000-dalton protein from Haemophilus influenzae.
Deich RA, et al.
Journal of Bacteriology, 170(2), 489-498 (1988)

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service