Skip to Content
Merck
All Photos(3)

Documents

K0753

Sigma-Aldrich

Kinetin

suitable for plant cell culture, crystalline

Synonym(s):

6-Furfurylaminopurine, N6-Furfuryladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N5O
CAS Number:
Molecular Weight:
215.21
Beilstein:
21703
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

Assay

≥98% (TLC and HPLC)

Quality Level

form

crystalline

technique(s)

cell culture | plant: suitable

color

white

mp

264-270 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccco3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetin is an artificial cytokinin. It is named due to its ability to stimulate cell division.

Application

Kinetin has been used:
  • in modified Murashige and Skoog (MS) media for shoot initiation(4)
  • as plant growth regulator (PGR) for shoot induction and proliferation(5)
  • to study its effects on the growth of two chlorophyte Microalgae, Haematococcus pluvialis and Dunaliella salina(6)

Biochem/physiol Actions

Kinetin is an adenine-type cytokinin phytohormone that is used in plant culture media such as Murashige & Skoog media in conjunction with auxins such as indole-3-acetic acid (IAA), indole-3-butyric acid (IBA), 1-napthaleneacetic acid (NAA) or others to induce the fomation of callus and to regenerate plant tissues from callus.
Kinetin is capable of inducing the opening of grass stomata.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hormonal regulation of ion transporters: the guard cell system
New comprehensive biochemistry (1999)
Hormones in Growth and Development
Reference Module in Life Sciences (2017)
Hugo A Méndez-Hernández et al.
Plants (Basel, Switzerland), 10(10) (2021-10-24)
Auxin plays a central role in growth and plant development. To maintain auxin homeostasis, biological processes such as biosynthesis, transport, degradation, and reversible conjugation are essential. The Gretchen Hagen 3 (GH3) family genes codify for the enzymes that esterify indole-3-acetic
Gabriel L Fiorin et al.
Bio-protocol, 10(8), e3588-e3588 (2021-03-05)
Plants recognize a wide variety of microbial molecules to detect and respond to potential invaders. Recognition of Microbe-Associated Molecular Patterns (MAMPs) by cell surface receptors initiate a cascade of biochemical responses that include, among others, ion fluxes across the plasma
Yanmei Chen et al.
The Plant journal : for cell and molecular biology, 63(1), 1-17 (2010-04-09)
Protein phosphorylation/dephosphorylation is a central post-translational modification in plant hormone signaling, but little is known about its extent and function. Although pertinent protein kinases and phosphatases have been predicted and identified for a variety of hormone responses, classical biochemical approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service