Skip to Content
Merck
All Photos(1)

Documents

B3274

Sigma-Aldrich

6-Benzylaminopurine solution

1 mg/mL, suitable for plant cell culture

Synonym(s):

BAP, BA, N6-Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

sterility

sterile-filtered

Quality Level

form

solution

concentration

1 mg/mL

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

General description

6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin.

Application

6-Benzylaminopurine solution has been used as a component in the Murashige & Skoog medium (MS) for culturing mandarin explants and plantlets of Dendrocalamus asper (Schultes f.). It has also been used as a supplement in Nitsch and Nitsch medium (NN) of grapevine explants,

Biochem/physiol Actions

6-Benzylaminopurine (BAP) is used as a cytokinin supplement to plant growth medium It increases post-harvest life of green vegetables by preventing chlorophyll degradation. It is used in micropropagation. However, in some plants, BAP leads to inhibition in growth and root formation. BAP phenyl ring based derivatives are widely used to test their anti-cancer effects

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is BA (6-benzyladenine) BAP (6-benzylaminopurine)?
da Silva JAT
The Asian and Australasian journal of Plant Science and Biotechnology , 6(1), 121-124 (2012)
Somatic embryogenesis of Vitis vinifera L.(cv. Sugraone) from stigma and style culture
Morgana C, et al.
Vitis, 43(4), 169-173 (2004)
Karel Dolezal et al.
Bioorganic & medicinal chemistry, 14(3), 875-884 (2005-10-11)
To study the structure-activity relationships of aromatic cytokinins, the cytokinin activity at both the receptor and cellular levels, as well as CDK inhibitory and anticancer properties of 38 6-benzylaminopurine (BAP) derivatives were compared in various in vitro assays. The compounds
Elimination of Citrus psorosis virus by somatic embryogenesis from stigma and style cultures
Donghia AM, et al.
Plant Pathology, 50(2), 266-269 (2001)
6-Benzylaminopurine improves shelf life, organoleptic quality and healthpromoting compounds of fresh-cut broccoli florets
Siddiqui MW, et al.
J. Sci. Ind. Res., 70(461), 461-465 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service