Skip to Content
Merck
All Photos(1)

Key Documents

I6154

Sigma-Aldrich

ProteoMass Insulin chain B oxidized MALDI-MS Standard

vial of 10 nmol

Synonym(s):

Insulin Chain B Oxidized from bovine pancreas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C157H232N40O47S2
CAS Number:
Molecular Weight:
3495.89
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.24
Pricing and availability is not currently available.

mol wt

monoisotopic mol wt 3,493.6513 Da

Quality Level

packaging

vial of 10 nmol

format

neat

SMILES string

[S](=O)(=O)(O)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)Cc8c

InChI

1S/C157H232N40O47S2/c1-79(2)57-104(181-130(211)86(15)172-136(217)103(50-53-125(209)210)180-152(233)127(84(11)12)194-148(229)107(60-82(7)8)184-145(226)113(67-95-70-165-78-171-95)189-150(231)115(74-198)176-123(206)73-169-133(214)116(75-245(239,240)241)191-140(221)105(58-80(3)4)182-144(225)112(66-94-69-164-77-170-94)188-138(219)102(48-51-119(160)202)178-146(227)114(68-120(161)203)190-153(234)126(83(9)10)193-131(212)98(159)61-89-31-21-18-22-32-89)139(220)185-110(64-92-40-44-96(200)45-41-92)142(223)183-106(59-81(5)6)147(228)195-128(85(13)14)154(235)192-117(76-246(242,243)244)134(215)168-71-121(204)174-101(49-52-124(207)208)137(218)177-99(38-29-55-166-157(162)163)132(213)167-72-122(205)175-108(62-90-33-23-19-24-34-90)141(222)186-109(63-91-35-25-20-26-36-91)143(224)187-111(65-93-42-46-97(201)47-43-93)149(230)196-129(88(17)199)155(236)197-56-30-39-118(197)151(232)179-100(37-27-28-54-158)135(216)173-87(16)156(237)238/h18-26,31-36,40-47,69-70,77-88,98-118,126-129,198-201H,27-30,37-39,48-68,71-76,158-159H2,1-17H3,(H2,160,202)(H2,161,203)(H,164,170)(H,165,171)(H,167,213)(H,168,215)(H,169,214)(H,172,217)(H,173,216)(H,174,204)(H,175,205)(H,176,206)(H,177,218)(H,178,227)(H,179,232)(H,180,233)(H,181,211)(H,182,225)(H,183,223)(H,184,226)(H,185,220)(H,186,222)(H,187,224)(H,188,219)(H,189,231)(H,190,234)(H,191,221)(H,192,235)(H,193,212)(H,194,229)(H,195,228)(H,196,230)(H,207,208)(H,209,210)(H,237,238)(H4,162,163,166)(H,239,240,241)(H,242,243,244)/t86-,87-,88+,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,126-,127-,128-,129-/m0/s1

InChI key

CZJQESISDHGFTC-DQCCOYHDSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Phe-Val-Asn-Gln-His-Leu-Cys(SO3H)-Gly-Ser-His-Leu-Val-Glu-Ala-Leu-Tyr-Leu-Val-Cys(SO3H)-Gly-Glu-Arg-Gly-Phe-Phe-Tyr-Thr-Pro-Lys-Ala

Application

Peptide calibration standard for MALDI-MS instruments.

Biochem/physiol Actions

Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids. It inhibits the breakdown of glycogen, protein, and fat. The β chain is a substrate for carboxypeptidase Y.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuchu Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(12), E2524-E2532 (2017-03-08)
Pain-producing animal venoms contain evolutionarily honed toxins that can be exploited to study and manipulate somatosensory and nociceptive signaling pathways. From a functional screen, we have identified a secreted phospholipase A2 (sPLA2)-like protein, BomoTx, from the Brazilian lancehead pit viper

Articles

One of the most important aspects of our ultra-pure MALDI matrix substances is their ability to dissolve rapidly and completely; a brief vortex mixing is typically sufficient.

Related Content

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service