Skip to Content
Merck
All Photos(1)

Key Documents

56197

Supelco

3-Hydroxypicolinic acid

matrix substance for MALDI-MS, ≥99.0% (HPLC)

Synonym(s):

3-HPA, 3-Hydroxypyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
Beilstein:
118954
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (HPLC)

form

powder or crystals

analyte chemical class(es)

oligonucleotides, oligosaccharides

technique(s)

MALDI-MS: suitable

mp

213-218 °C (lit.)

cation traces

Ba: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤20 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤50 mg/kg

SMILES string

OC(=O)c1ncccc1O

InChI

1S/C6H5NO3/c8-4-2-1-3-7-5(4)6(9)10/h1-3,8H,(H,9,10)

InChI key

BRARRAHGNDUELT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Hydroxypicolinic acid (3-HPA) is a matrix material used in matrix-assisted laser desorption/ionization (MALDI) based mass spectroscopy (MS) for the analysis of large biomolecules and quantification of oligonucleotides.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

3-Hydroxypicolinic acid has been used in the study conducted to isolate and characterize bacteria degrading-quinoline in subsurface sediments.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase diagnostics and LIF-imaging of 3-hydroxypicolinic acid MALDI-matrix plumes
Puretzky AA and Geohegan DB
Chemical Physics Letters, 286(5-6), 425-432 (1998)
P Juhasz et al.
Carbohydrate research, 270(2), 131-147 (1995-04-30)
Molecular weights of heparin-derived oligosaccharides ranging from disaccharides to hexadecasaccharides have been determined by matrix-assisted laser desorption ionization time-of-flight mass spectrometry. While these compounds ionize poorly or not at all when used as such, a strong signal can be obtained
K Tang et al.
Nucleic acids research, 23(16), 3126-3131 (1995-08-25)
Matrix-assisted laser desorption/ionization (MALDI) time-of-flight (TOF) mass spectrometry was used to analyze short DNA duplex probes with one strand immobilized on solid supports (straptavidin-coated magnetic beads or controlled pore glass beads). Only the non-immobilized strand could be detected. Partial denaturation
H Suzuki et al.
Analytical chemistry, 69(22), 4554-4559 (1998-01-07)
Matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS), has been used for structure characterization of 1-aminopyrene-3,6,8-trisulfonate (APTS)-derivatized oligosaccharides previously separated by capillary electrophoresis (CE). The resolved components were first isolated by employing an automated high-resolution fraction collector. Using on-probe sample
Isolation and characterization of quinoline-degrading bacteria from subsurface sediments.
Applied and Environmental Microbiology, 55 (4), 1029-1032 (1989)

Related Content

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

ZipTip® micro-SPE pipette tips are used as a single-step desalting, concentration, and purification tool for complex samples before mass spec analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service