Skip to Content
Merck
All Photos(1)

Key Documents

HT5014

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

case of 24 × 120 mL, histological tissue fixative

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.47

packaging

case of 24 × 120 mL

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formalin solution, neutral buffered, 10% has been used to fix skin samples, tumor samples and HepG2 human liver carcinoma cells.
Popular general purpose histological fixative.

Packaging

Case containers are polypropylene with white leak-proof lids and are partially prefilled with 10% NBF.

Physical form

Neutral buffered solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclin D3 deficiency inhibits skin tumor development, but does not affect normal keratinocyte proliferation.
Lee S H, et al.
Oncology Letters, 14(3), 2723-2734 (2017)
Mostafa El-Diwany et al.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS, 70(9), 1210-1217 (2017-07-09)
The most common complications of irradiated implant-based mammary reconstruction are fibrosis and capsular contracture. The indications for postmastectomy adjuvant radiotherapy have considerably broadened. Facing an increased number of patients who will require radiotherapy, most guidelines recommend delaying reconstruction after radiotherapy
Catalase inhibition induces pexophagy through ROS accumulation.
Lee J N, et al.
Biochemical and Biophysical Research Communications, 501(3), 696-702 (2018)
Anna Maria D'Erchia et al.
Scientific reports, 7(1), 10046-10046 (2017-09-01)
ALS is a devastating and debilitating human disease characterized by the progressive death of upper and lower motor neurons. Although much effort has been made to elucidate molecular determinants underlying the onset and progression of the disorder, the causes of
Fumihiko Namba et al.
PloS one, 9(3), e90936-e90936 (2014-03-07)
Premature infants exposed to hyperoxia suffer acute and long-term pulmonary consequences. Nevertheless, neonates survive hyperoxia better than adults. The factors contributing to neonatal hyperoxic tolerance are not fully elucidated. In contrast to adults, heme oxygenase (HO)-1, an endoplasmic reticulum (ER)-anchored

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service