Skip to Content
Merck
All Photos(1)

Key Documents

HT501320

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

histological tissue fixative

Synonym(s):

Formaldehyde, Methanal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

synthetic

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formalin solution, neutral buffered, 10% is the most commonly used fixative for light microscopy. It requires a relatively short period of fixation. It is suitable for the long-term storage of tissue. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining.

Application

Formalin solution, neutral buffered, 10% has been used as a popular general purpose histological fixative to fix tissues.[1][2][3]
Popular general purpose histological fixative.

Packaging

Item is packaged in a 10L Cubitainer consisting of a plastic bladder inside of a cardboard box with an attached spigot.

Physical form

Neutral buffered solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of partially and fully chemically-modified siRNA in conjugate-mediated delivery in vivo.
Hassler M R, et al.
Nucleic Acids Research, 46(5), 2185-2196 (2018)
Quantifying size and number of adipocytes in adipose tissue.
Parlee S D, et al.
Methods in Enzymology, 537, 93-122 (2014)
Recapitulating spatiotemporal tumor heterogeneity in vitro through engineered breast cancer microtissues.
Mazio C, et al.
Acta Biomaterialia, 73, 236-249 (2018)
Ted M Hsu et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(48), 30744-30754 (2020-11-18)
Thirst is a highly potent drive that motivates organisms to seek out and consume balance-restoring stimuli. The detection of dehydration is well understood and involves signals of peripheral origin and the sampling of internal milieu by first order homeostatic neurons
Matteo Riva et al.
Biology open, 8(9) (2019-09-13)
Recently, several promising treatments for high-grade gliomas (HGGs) failed to provide significant benefit when translated from the preclinical setting to patients. Improving the animal models is fundamental to overcoming this translational gap. To address this need, we developed and comprehensively

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service