Skip to Content
Merck
All Photos(2)

Key Documents

HT110332

Sigma-Aldrich

Eosin Y Solution, Alcoholic, with Phloxine

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christy L Trejo et al.
PloS one, 14(2), e0212031-e0212031 (2019-02-23)
We describe the use of a ligation-based targeted whole transcriptome expression profiling assay, TempO-Seq, to profile formalin-fixed paraffin-embedded (FFPE) tissue, including H&E stained FFPE tissue, by directly lysing tissue scraped from slides without extracting RNA or converting the RNA to
Yael Aylon et al.
Genes & development, 30(7), 786-797 (2016-03-26)
The Hippo signaling pathway is a major regulator of organ size. In the liver, Hippo pathway deregulation promotes hyperplasia and hepatocellular carcinoma primarily through hyperactivation of its downstream effector, YAP. The LATS2 tumor suppressor is a core member of the
Insu Kwon et al.
The journal of physiological sciences : JPS, 68(3), 269-280 (2017-02-19)
Elevation of anabolism and concurrent suppression of catabolism are critical metabolic adaptations for muscular hypertrophy in response to resistance exercise (RE). Here, we investigated if RE-induced muscular hypertrophy is acquired by modulating a critical catabolic process autophagy. Male Wistar Hannover
Su Ji Bae et al.
Molecules (Basel, Switzerland), 25(11) (2020-06-12)
Perilla oil has been considered to have excellent potential for treating various diseases due to its contents of beneficial fatty acids, such as α-linolenic acid, oleic acid and linoleic acid. The therapeutic effects and molecular mechanism of an α-linolenic acid-enriched
Mi Rim Lee et al.
Experimental and therapeutic medicine, 17(3), 2185-2193 (2019-03-15)
The therapeutic effects of mulberry (Morus alba) leaves on lipid metabolism, including lipogenesis, lipolysis and hyperlipidemia are widely known, although their fermented products are yet to be applied. To investigate the therapeutic effects of a novel extract of mulberry leaves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service